Skip to Content
Merck
All Photos(1)

Key Documents

279897

Sigma-Aldrich

(R)-(−)-2-Phenylpropionic acid

97%

Synonym(s):

(R)-(−)-Hydratropic acid, (R)-HTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H5)CO2H
CAS Number:
Molecular Weight:
150.17
Beilstein:
2207688
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D −72°, c = 1.6 in chloroform

optical purity

ee: 98% (HPLC)

refractive index

n20/D 1.523 (lit.)

bp

115 °C/1 mmHg (lit.)

mp

29-30 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

C[C@@H](C(O)=O)c1ccccc1

InChI

1S/C9H10O2/c1-7(9(10)11)8-5-3-2-4-6-8/h2-7H,1H3,(H,10,11)/t7-/m1/s1

InChI key

YPGCWEMNNLXISK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block. Resolving agent

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christiane Kiske et al.
Journal of agricultural and food chemistry, 67(4), 1187-1196 (2019-01-04)
The enantiomers of a homologous series (C6-C10) of 2-mercapto-4-alkanones were obtained by lipase-catalyzed kinetic resolution of the corresponding racemic 2-acetylthio-4-alkanones. Their configurations were assigned via vibrational circular dichroism and 1H NMR anisotropy based methods. Odor thresholds and odor qualities were
Chunze Li et al.
The Journal of pharmacology and experimental therapeutics, 305(1), 250-256 (2003-03-22)
Two alternative metabolic pathways, acyl glucuronidation and acyl-CoA formation, are implicated in the generation of reactive acylating metabolites of carboxylic acids. Here, we describe studies that determine the relative importance of these two pathways in the metabolic activation of a
M J Thomason et al.
The Journal of pharmacy and pharmacology, 49(3), 263-269 (1997-03-01)
Previous studies have demonstrated that Verticillium lecanii might be used as a microbial model of the inversion of 2-arylpropionic acids in man. This paper describes the optimization of the inversion process in respect of culture medium, pH, cell density and
Chunze Li et al.
Chemical research in toxicology, 15(11), 1480-1487 (2002-11-20)
A series of studies was conducted to investigate the potential of (R)- and (S)-2-phenylpropionic acid (2-PPA) to undergo enantioselective covalent binding to protein in freshly isolated rat hepatocytes and to determine whether such covalent binding is dependent on acyl glucuronidation
Y Nakamura et al.
Drug metabolism and disposition: the biological fate of chemicals, 15(4), 529-534 (1987-07-01)
Optical isomerization of 2-phenylpropionic acid (hydratropic acid, HTA) was studied in the organs of male rat in vitro. (R)-(-)-HTA was not isomerized by rat liver homogenate even after the addition of CoA, ATP, and Mg2+ to the incubation mixture; however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service