Skip to Content
Merck
All Photos(1)

Documents

154210

Sigma-Aldrich

(R)-(−)-Mandelic acid

ReagentPlus®, ≥99%

Synonym(s):

(R)-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2691094
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

≥99%

form

solid

optical activity

[α]23/D −153°, c = 2.5 in H2O

mp

131-133 °C (lit.)

SMILES string

O[C@@H](C(O)=O)c1ccccc1

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1

InChI key

IWYDHOAUDWTVEP-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(-)-Mandelic acid, a chiral resolving agent, is also used as a building block to synthesize pharmaceutical drugs such as penicillin and cephalosporin. It can be synthesized from (R,S)-mandelonitrile with high yield and enantioselectivity using nitrilase enzyme .

(R)-(−)-Mandelic acid is a carboxylic acid used as a starting material to synthesize antibiotics, antitumor agents, and antiobesity drugs.

Application

(R)-(-)-Mandelic acid has been used in studies to assess its ability to undergo spontaneous oscillatory chiral conversion and spontaneous condensation to form polymandelic acid.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of R-(-)-mandelic acid from mandelonitrile by Alcaligenes faecalis ATCC 8750.
Yamamoto K, et al.
Applied and Environmental Microbiology, 57(10), 3028-3032 (1991)
On the spontaneous condensation of selected hydroxy acids.
Sajewicz M, et al.
Acta Chromatographica , 21(2), 259-271 (2009)
Green synthesis aspects of (R)-(-)-mandelic acid; a potent pharmaceutically active agent and its future prospects
Rahul Vikram S et al.
Critical Reviews in Biotechnology, 43, 1226-1235 (2023)
Enzyme-catalysed optical resolution of mandelic acid via RS (?)-methyl mandelate in non-aqueous media.
Yadav GD and Sivakumar P.
Biochemical Engineering Journal, 19(2), 101-107 (2004)
TLC in a search for structural limitations of spontaneous oscillatory in-vitro chiral conversion. a-hydroxybutyric and mandelic acids.
Sajewicz M, et al.
J. Planar Chromatogr., 22(4), 241-248 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service