Skip to Content
Merck
All Photos(1)

Documents

274992

Sigma-Aldrich

O-Ethylhydroxylamine hydrochloride

97%

Synonym(s):

Ethoxyamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2ONH2 · HCl
CAS Number:
Molecular Weight:
97.54
Beilstein:
3606639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

130-133 °C (lit.)

SMILES string

Cl[H].CCON

InChI

1S/C2H7NO.ClH/c1-2-4-3;/h2-3H2,1H3;1H

InChI key

NUXCOKIYARRTDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

O-Ethylhydroxylamine hydrochloride was used in the determination of α-hydroxycarbonyl compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of α-hydroxycarbonyl and α-dicarbonyl compounds during degradation of monosaccharides.
NOVOtNy O, et al.
Czech Journal of Food Sciences, 25(3), 119-130 (2007)
Bo He et al.
Nature communications, 12(1), 4249-4249 (2021-07-14)
5-Hydroxymethylcytosine (5hmC) is an important epigenetic mark that regulates gene expression. Charting the landscape of 5hmC in human tissues is fundamental to understanding its regulatory functions. Here, we systematically profiled the whole-genome 5hmC landscape at single-base resolution for 19 types
Yafen Wang et al.
Chemical science, 10(2), 447-452 (2019-02-13)
5-Hydroxymethylcytosine (5hmC) is known as one of the vital players in nuclear reprogramming and the process of active DNA demethylation. Although the development of whole-genome sequencing methods for modified cytosine bases has burgeoned, the easily operated gene-specific loci detection of
Enkhtuul Tsogtbaatar et al.
Journal of experimental botany, 66(14), 4267-4277 (2015-02-26)
Pennycress (Thlaspi arvense L.), a plant naturalized to North America, accumulates high levels of erucic acid in its seeds, which makes it a promising biodiesel and industrial crop. The main carbon sinks in pennycress embryos were found to be proteins
Teresa Mairinger et al.
Analytical chemistry, 87(23), 11792-11802 (2015-10-30)
For the first time an analytical work flow based on accurate mass gas chromatography-quadrupole time-of-flight mass spectrometry (GC-QTOFMS) with chemical ionization for analysis providing a comprehensive picture of (13)C distribution along the primary metabolism is elaborated. The method provides a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service