270296
Calcium carbide
pieces, thickness <10 mm , typically, technical grade, ~80%
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CaC2
CAS Number:
Molecular Weight:
64.10
Beilstein:
3909011
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
technical grade
Assay
~80%
form
pieces
thickness
<10 mm , typically
density
2.22 g/mL at 25 °C (lit.)
SMILES string
[Ca++].[C-]#[C-]
InChI
1S/C2.Ca/c1-2;/q-2;+2
InChI key
UIXRSLJINYRGFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 1
Target Organs
Respiratory system
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
H Kjuus et al.
British journal of industrial medicine, 43(4), 237-242 (1986-04-01)
The overall mortality and the incidence of cancer have been studied among male employees at a plant producing calcium carbide. The cohort was defined as all men employed at the plant for at least 18 months in the period 1953
P Boffetta et al.
Cancer causes & control : CCC, 8(3), 444-472 (1997-05-01)
Epidemiologic evidence on the relationship between polycyclic aromatic hydrocarbons (PAH) and cancer is reviewed. High occupational exposure to PAHs occurs in several industries and occupations. Covered here are aluminum production, coal gasification, coke production, iron and steel foundries, tar distillation
G Malla et al.
Chemosphere, 58(2), 141-147 (2004-12-02)
Mitigation of methane (CH4) and nitrous oxide (N2O) emissions from soil is important to reduce the global warming. Efficacy of five nitrification inhibitors, i.e. neem (Azadirachta melia) cake, thiosulphate, coated calcium carbide, neem oil coated urea and dicyandiamide (DCD) and
Xiaoguang Jiao et al.
Science in China. Series C, Life sciences, 48 Suppl 1, 26-32 (2005-08-11)
A field experiment was carried out at the Shenyang Experimental Station of Ecology (CAS) in order to study the effects of slow-release urea fertilizers high polymer-coated urea (SRU1), SRU1 mixed with dicyandiamide DCD (SRU2), and SRU1 mixed with calcium carbide
Production of calcium carbide from fine biochars.
Guodong Li et al.
Angewandte Chemie (International ed. in English), 49(45), 8480-8483 (2010-09-14)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service