Skip to Content
Merck
All Photos(1)

Key Documents

268119

Sigma-Aldrich

Diethyl methylphosphonate

97%

Synonym(s):

Bis(diethoxyphosphinyl)methane, DEMP, Diethoxymethylphosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(O)(OC2H5)2
CAS Number:
Molecular Weight:
152.13
Beilstein:
1753416
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.414 (lit.)

bp

194 °C (lit.)

density

1.041 g/mL at 25 °C (lit.)

functional group

phosphonate

SMILES string

CCOP(C)(=O)OCC

InChI

1S/C5H13O3P/c1-4-7-9(3,6)8-5-2/h4-5H2,1-3H3

InChI key

NYYLZXREFNYPKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for the synthesis of:
  • Fluoroalkyl α- and β-aminophosphonates
  • Acyclic nucleoside phosphonates with branched phosphonoethoxy-Et chains as potential antivirals
  • Difluoromethyl arylphosphonates
  • Pyridone alkaloids with neuritogenic activity
  • Lipophilic meropenem-derived prodrugs
  • Phomactin cyclohexane core and diterpenoid framework
  • Phosphonylated peptides via solid-phase synthesis
Reagent employed in the synthesis of α-alkenyl, γ-hydroxyl, and dialkyl fluoroalkynyl phosphonates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 5779-5779 (1993)
Synthetic Communications, 21, 2341-2341 (1991)
Journal of the Chemical Society. Perkin Transactions 1, 2153-2153 (1993)
Ying Zhang et al.
Biomaterials, 26(33), 6736-6742 (2005-06-07)
Novel BAB type amphiphilic triblock copolymers consisting of poly (ethylene glycol) (PEG) (B) as hydrophilic segment and poly (epsilon-caprolactone) (PCL) (A) as hydrophobic block were prepared by coupling reaction using L-lysine methyl ester diisocyanate (LDI) as the chain extender. The
E Maurelli et al.
Free radical biology & medicine, 27(1-2), 34-41 (1999-08-12)
The spin trap 5-(diethoxyphosphoryl)-5-methyl-1-pyrroline N-oxide (DEPMPO) is an improved ESR probe to assess superoxide (O2*-) formation in the postischemic heart. We recently found that DEPMPO pretreatment improves recovery of cardiac function with the concomitant inhibition of postischemic O2*- production. By

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service