Skip to Content
Merck
All Photos(1)

Documents

241024

Sigma-Aldrich

Trimethyl phosphate

≥99%

Synonym(s):

TMP, TMPA, TMPO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3PO
CAS Number:
Molecular Weight:
140.07
Beilstein:
1071731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.395 (lit.)

bp

197 °C (lit.)

mp

−46 °C (lit.)

density

1.197 g/mL at 25 °C (lit.)

SMILES string

COP(=O)(OC)OC

InChI

1S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3

InChI key

WVLBCYQITXONBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The trimethyl phosphate levels in environmental water was determined by direct analysis real-time ionization source interfaced with a triple quadrupole mass spectrometer (DART-MS/MS) with multiple reaction monitoring (MRM).

Application


  • Enhanced Battery Performance: Trimethyl phosphate is utilized to manipulate the ionic conductivity and interfacial compatibility in polymer-in-dual-salt electrolytes, significantly extending the operable temperature range of quasi-solid metal batteries (Lin et al., 2024).

  • Thermochemical Properties Analysis: Research on trimethyl phosphate′s thermochemistry properties and rate kinetics, particularly H-Atom Abstraction reactions, provides valuable insights into its stability and reactivity, which are crucial for various chemical synthesis applications (Bruce and Li, 2023).

  • Organophosphorus Flame Retardants Detection: Trimethyl phosphate is explored for its role in the screening of organophosphorus flame retardant residues in food samples, specifically rice, utilizing advanced chromatographic techniques to ensure food safety and compliance with environmental regulations (Li et al., 2023).

  • Interfacial Reaction Regulation in Batteries: The study on regulating interfacial reactions through electrolyte chemistry, involving trimethyl phosphate, highlights its potential to create gradient interphases for low-temperature zinc metal batteries, contributing to enhanced battery safety and efficiency (Wang et al., 2023).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaowei Wang et al.
Journal of chromatography. A, 1333, 134-137 (2014-02-20)
Trimethyl phosphate (TMP) is used extensively in industrial chemical processes. Due to the high polarity and volatility, methods for its quantification in environmental samples have not been well developed. Currently, the pollution status of TMP in the environment still has
Marianna Ambrico et al.
Physical chemistry chemical physics : PCCP, 21(23), 12380-12388 (2019-05-30)
Nanoscale disassembly of mussel-inspired polydopamine (PDA) in ionic liquids (ILs) was recently shown to induce an electron paramagnetic resonance (EPR)-detectable reorganization of free radical centers in the resulting nanoparticles (NPs) in an IL-controlled manner. Herein, we report electrical impedance spectroscopy
R V Banerjee et al.
Biochemistry, 27(25), 9062-9070 (1988-12-13)
The transfer of 17O and/or 18O from (COOH-17O or -18O) enriched substrates to inorganic phosphate (Pi) has been demonstrated for two enzyme-catalyzed reactions involved in folate biosynthesis and glutamylation. COOH-18O-labeled folate, methotrexate, and dihydropteroate, in addition to [17O]-glutamate, were synthesized
Qingguo Meng et al.
Environmental science & technology, 45(7), 3000-3005 (2011-03-03)
Due to the neurotoxicity of organophosphate (OP) pesticides and nerve agents synthesized as military or terror agents, their safe destruction and disposal is of considerable current importance. A representative OP, trimethyl phosphate (TMP), was adsorbed onto NaX zeolite, two mesoporous
K Sundararajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(10), 1855-1867 (2000-09-16)
Trimethyl phosphate (TMP) and acetylene were codeposited in nitrogen and argon matrices and adducts of these species were identified using infrared spectroscopy. Formation of the adducts was evidenced by shifts in the vibrational frequencies of the modes involving the TMP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service