Skip to Content
Merck
All Photos(1)

Key Documents

268011

Sigma-Aldrich

Ethyl oleate

98%

Synonym(s):

Oleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOC2H5
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727318
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

216-218 °C/15 mmHg

mp

−32 °C (lit.)

solubility

chloroform: soluble 10%

density

0.87 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-

InChI key

LVGKNOAMLMIIKO-QXMHVHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl oleate can be used as a starting material to synthesize stearic acid hydrazide intermediate, which can undergo intermolecular cyclization reaction with various aliphatic acids and aromatic acids to yield 1,3,4-oxadiazole derivatives. It is also reduced to oleyl alcohol via Bouveault−Blanc reduction reaction.
Ethyl oleate was used to prepare the oily phase of self-microemulsifying drug delivery system (SMEDDS) for tacrolimus (Tac).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian S Bodnar et al.
The Journal of organic chemistry, 74(6), 2598-2600 (2009-02-18)
Significantly improved Bouveault-Blanc conditions for ester reduction have been developed using sodium in silica gel (Na-SG), a free-flowing powder that can be easily handled in the open atmosphere. Primary alcohols were prepared in excellent yield from a variety of aliphatic
Kalidas Ramamoorthy et al.
American journal of physiology. Gastrointestinal and liver physiology, 321(2), G123-G133 (2021-06-03)
Vitamin B7 (biotin) is essential for normal health and its deficiency/suboptimal levels occur in a variety of conditions including chronic alcoholism. Mammals, including humans, obtain biotin from diet and gut-microbiota via absorption along the intestinal tract. The absorption process is
Marietta von Suesskind-Schwendi et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 67(6), 469-473 (2013-05-15)
This study evaluated the pharmacokinetics of tacrolimus (Tac) in a novel self-microemulsifying drug delivery system (SMEDDS) for improved oral administration. SMEDDS Tac consisted of ethyl oleate as the oily phase, Solutol HS 15 as the surfactant and glycofurol as the
Michael D Hardy et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8187-8192 (2015-06-24)
Cell membranes are dynamic structures found in all living organisms. There have been numerous constructs that model phospholipid membranes. However, unlike natural membranes, these biomimetic systems cannot sustain growth owing to an inability to replenish phospholipid-synthesizing catalysts. Here we report
Aihua Yu et al.
Journal of pharmaceutical sciences, 100(3), 933-941 (2010-09-24)
The main objective of the study was to investigate the efficacy of microemulsion (ME) to facilitate bioavailability of puerarin (PUE) after oral and nasal administration. The pseudo-ternary phase diagrams were constructed to screen the ME components and optimize the ME

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service