Skip to Content
Merck
All Photos(2)

Key Documents

263303

Sigma-Aldrich

Tellurium

granular, −5-+50 mesh, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Te
CAS Number:
Molecular Weight:
127.60
EC Number:
MDL number:
UNSPSC Code:
12141913
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

granular

resistivity

5.8-33 μΩ-cm, 20°C

particle size

−5-+50 mesh

bp

990 °C (lit.)

mp

450 °C (lit.)

density

6.24 g/mL at 25 °C (lit.)

SMILES string

[Te]

InChI

1S/Te

InChI key

PORWMNRCUJJQNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 4 - Lact. - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hridyesh Kumar et al.
Carbohydrate polymers, 97(2), 327-334 (2013-08-06)
The present work describes synthesis of water soluble L-cysteine-functionalized CdTe quantum dots (QDs) with size tunable emission at different time intervals for chitosan based film. The characterization of the synthesized CdTe QDs-chitosan film was made by Fourier transform infrared (FTIR)
E C Stangherlin et al.
Toxicology, 207(2), 231-239 (2004-12-15)
The effect of single maternal subcutaneous (s.c.) injection of 0.12 mg/kg diphenyl ditelluride, (PhTe)2, diluted in canola oil at days 6, 10 or 17 of gestation were evaluated in Wistar rats. The reduction of body weight gain was statistically significant
Edward R T Tiekink
Dalton transactions (Cambridge, England : 2003), 41(21), 6390-6395 (2012-01-19)
Despite being disparaged for their malodorous and toxic demeanour, compounds of selenium, a bio-essential element, and tellurium, offer possibilities as therapeutic agents. Herein, their potential use as drugs, for example, as anti-viral, anti-microbial, anti-inflammatory agents, etc., will be surveyed along
Katja C Nowack et al.
Nature materials, 12(9), 787-791 (2013-06-19)
The quantum spin Hall (QSH) state is a state of matter characterized by a non-trivial topology of its band structure, and associated conducting edge channels. The QSH state was predicted and experimentally demonstrated to be realized in HgTe quantum wells.
Guizhong Wang et al.
Optics express, 21(4), 4703-4708 (2013-03-14)
The octave-spanning spectrum was generated in a tellurite glass based microstructured fiber pumped by a 528 MHz repetition rate Yb:fiber ring laser without amplification. The laser achieved 40% output optical-to-optical efficiency with the output power of 410 mW. By adjusting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service