Skip to Content
Merck
All Photos(2)

Key Documents

255645

Sigma-Aldrich

Cesium carbonate

99.995% trace metals basis

Synonym(s):

Carbonic acid dicesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2CO3
CAS Number:
Molecular Weight:
325.82
Beilstein:
4546405
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.995% trace metals basis

form

powder

mp

610 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

[Cs+].[Cs+].[O-]C([O-])=O

InChI

1S/CH2O3.2Cs/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

FJDQFPXHSGXQBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cesium carbonate is a mild inorganic base widely used inelectroluminescent devices and organic synthesis. Due to its excellent electroninjection capability, it is used as an interface modifying material in organiclight-emitting diodes (OLED), organic field effect transistors (OFET), organicsolar cells (OSCs), and perovskite solar cells.

Application

Cesium carbonate can be used as:
  • A cathode buffer layer to enhance device stability and power conversion efficiency in polymer solar cells.
  • A precursor to prepare alkali-based electrolyte for selective reduction of CO2 by electrocatalysis.
  • A base in many organic transformations such as C, N, O alkylation, and arylation reactions.
Promotes the efficient O-alkylation of alcohols to form mixed alkyl carbonates.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney,Adrenal gland,Testes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Wang et al.
ChemSusChem, 14(4), 1131-1139 (2021-01-08)
Photocatalytic reduction of CO2 into value-added chemical fuels is an appealing approach to address energy crisis and global warming. CsPbBr3 quantum dots (QDs) are good candidates for CO2 reduction because of their excellent photoelectric properties, including high molar extinction coefficient
Xiangtong Zhang et al.
Journal of materials chemistry. C, 6(37), 10101-10105 (2018-12-07)
Lead halide perovskite nanocrystals (NCs) exhibit excellent tunable emissions covering the entire visible spectral region, but they do not emit near-infrared (NIR) light. We synthesized rare earth element doped CsPbCl3 NCs for NIR emission. The Yb3+ doped CsPbCl3 NCs emitted
Chu, F. et al.
Tetrahedron Letters, 40, 1847-1847 (1999)
Aanindeeta Banerjee et al.
Nature, 531(7593), 215-219 (2016-03-11)
Using carbon dioxide (CO2) as a feedstock for commodity synthesis is an attractive means of reducing greenhouse gas emissions and a possible stepping-stone towards renewable synthetic fuels. A major impediment to synthesizing compounds from CO2 is the difficulty of forming
Yanxiu Li et al.
ACS nano, 13(7), 8237-8245 (2019-07-12)
Semiconductor nanorods (NRs) offer the useful property of linearly polarized light emission. While this would be an attractive functionality for strongly emitting perovskite nanoparticles, to date, there has been limited success in demonstrating a direct chemical synthesis of cesium lead

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service