Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cesium carbonate
Synonym(s):
Carbonic acid dicesium
Linear Formula:
Cs
2
CO
3
CAS Number:
534-17-8
Molecular Weight:
325.82
EC Number:
208-591-9
Products
Product Category
inorganic bases (5)
salts (1)
Brand
Sigma-Aldrich (7)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Puriss (1)
Puriss P.A. (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
441902
Cesium carbonate,
ReagentPlus
®
, 99%
Expand
View Pricing
202126
Cesium carbonate,
99.9% trace metals basis
Expand
View Pricing
255645
Cesium carbonate,
99.995% trace metals basis
Expand
View Pricing
554855
Cesium carbonate,
99.95% trace metals basis
Expand
View Pricing
20959
Cesium carbonate,
puriss. p.a., ≥99.0%
Expand
View Pricing
1.02040
Cesium carbonate,
extra pure, Technipur
®
Expand
View Pricing
20960
Cesium carbonate,
purum p.a., ≥98.0% (T)
Expand
View Pricing