Skip to Content
Merck
All Photos(4)

Key Documents

254274

Sigma-Aldrich

Lithium hydroxide monohydrate

greener alternative

99.95% trace metals basis

Synonym(s):

Lithine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOH · H2O
CAS Number:
Molecular Weight:
41.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

Quality Level

Assay

99.95% trace metals basis

form

crystalline

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤550.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1

InChI key

GLXDVVHUTZTUQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hydroxide monohydrate is a strongly alkaline crystalline solid soluble in water. It is prepared industrially by recovering it from spodumene or by evaporating a solution prepared by reacting calcium hydroxide and lithium carbonate. It is mainly used in the synthesis of cathode materials for lithium-ion batteries, such as lithium cobalt oxide (LiCoO2) and lithium iron phosphate. It is also preferred over lithium carbonate for producing lithium nickel manganese cobalt oxides.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium hydroxide monohydrate can be used mainly for the synthesis of cathode materials for Lithium-ion batteries:
  • To synthesize Olivine LiFePO4 nanoplates with different crystal orientations by glycol-based solvothermal process. It demonstrated that the crystal orientation of cathode nanoplates plays an important role in the performance.
  • As a precursor for the synthesis of LiFePO4 platelets under hydrothermal synthesis conditions. It exhibited homogeneous particle size distribution, greater electrochemical performances and cycle life, and morphology control.
  • To synthesize Ni-rich Li[NixCoyMn1–x–y]O2 gradient cathodes (NCM). These cathodes exhibited enhanced cycling and chemical stability due to their strong crystallographic texture, unique particle morphology, and highly correlated particle orientation.
  • As a precursor to prepare porous salt hydrate-based composites for low-temperature thermochemical heat storage.
  • As an electrolyte additive in lithium-ion batteries.
Our Lithium hydroxide monohydrate which is water soluble, with a purity of 99.95%, is an excellent choice as a source for Lithium to synthesize the cathode materials for Lithium-ion batteries. The high purity on trace metals basis makes it highly suitable for cathode formation, ensuring optimal electrochemical performance.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Energy Ni-Rich Li [Ni x Co y Mn1--x--y] O2 Cathodes via Compositional Partitioning for Next-Generation Electric Vehicles
Chong S Yoon, et al
Chemistry of Materials, 29, 10436-10445 (2017)
Viktoria V Sursyakova et al.
Electrophoresis, 39(8), 1079-1085 (2018-02-08)
Cyclodextrins (CD) form inclusion complexes with different "guests" owing to the fact that the shape of the CD molecule is a truncated cone with a hydrophobic cavity. The adducts of CD with metal complexes remain scantily explored. In this study
Mechanism for Hydrothermal Synthesis of LiFePO4 Platelets as Cathode Material for Lithium-Ion Batteries
Xue Qin, et al.
The Journal of Physical Chemistry C, 114, 16806-16812 (2010)
Crystal Orientation Tuning of LiFePO4 Nanoplates for High Rate Lithium Battery Cathode Materials
Li Wang, et al
Nano Letters, 12, 5632-5636 (2012)
Construction of biomass waste derived hierarchical porous biochar framework based lithium hydroxide composites for highly efficient and durable low temperature thermochemical heat storage
Xiangyu Yang, et al.
Journal of Cleaner Production, 359, 132047-132047 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service