Skip to Content
Merck
All Photos(1)

Key Documents

252239

Sigma-Aldrich

1-Chlorotetradecane

98%

Synonym(s):

Myristyl chloride, Tetradecyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13Cl
CAS Number:
Molecular Weight:
232.83
Beilstein:
1742639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

139-142 °C/4 mmHg (lit.)

density

0.859 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCl

InChI

1S/C14H29Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-14H2,1H3

InChI key

RNHWYOLIEJIAMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phase behavior and phase separation kinetics of ternary nonionic microemulsion system composed of pentaethylene glycol dodecyl ether, water and 1-chlorotetradecane was studied.

Application

1-Chlorotetradecane has been used:
  • in preparation of nonionic microemulsion of pentaoxyethylene glycol dodecyl ether, water and 1-chlorotetradecane
  • as internal standard in determination of 3-chloropropane-1,2-diol in hydrolyzed vegetable protein by capillary gas chromatography with electrolytic conductivity detection

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of 3-chloropropane-1, 2-diol in hydrolysed vegetable proteins by capillary gas chromatography with electrolytic conductivity detection.
Spyres G.
Journal of Chromatography A, 638(1), 71-74 (1993)
G Roshan Deen et al.
The journal of physical chemistry. B, 114(23), 7769-7776 (2010-05-25)
A clear and stable nonionic model microemulsion consisting of pentaoxyethylene glycol dodecyl ether (C(12)E(5)), water, and 1-chlorotetradecane (CLTD) was prepared. This system was subjected to a systematic temperature quench (perturbation out of equilibrium) in steps of 1.0 degrees C from
G Roshan Deen et al.
The journal of physical chemistry. B, 113(20), 7138-7146 (2009-05-15)
The phase behavior and phase separation kinetics of a model ternary nonionic microemulsion system composed of pentaethylene glycol dodecyl ether (C12E5), water, and 1-chlorotetradecane were studied. With increasing temperature, the microemulsion exhibits the following rich phase behavior: oil-in-water phase (L1+O)
Renu Sharma et al.
Physical chemistry chemical physics : PCCP, 17(36), 23582-23594 (2015-08-25)
The present study aims to develop an understanding of the interactions between an anionic polyelectrolyte, poly sodium 4-styrene sulphonate (NaPSS), and cationic surface active imidazolium based ionic liquids (SAILs), [Cnmim][Cl] (n = 10, 12, 14) using a multi-technique approach. Various
Tânia E Sintra et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 20(5), 727-735 (2019-01-24)
Twelve surface-active ionic liquids (SAILs) and surface-active derivatives, based on imidazolium, ammonium, and phosphonium cations and containing one, or more, long alkyl chains in the cation and/or the anion, were synthetized and characterized. The aggregation behavior of these SAILs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service