Skip to Content
Merck
All Photos(1)

Key Documents

274496

Sigma-Aldrich

1-Chloro-2,2-dimethylpropane

new

98%

Synonym(s):

Neopentyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2Cl
CAS Number:
Molecular Weight:
106.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.4039 (lit.)

bp

84-85 °C (lit.)

mp

−20 °C (lit.)

density

0.866 g/mL at 25 °C (lit.)

functional group

alkyl halide
chloro

SMILES string

CC(C)(C)CCl

InChI

1S/C5H11Cl/c1-5(2,3)4-6/h4H2,1-3H3

InChI key

JEKYMVBQWWZVHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Chloro-2,2-dimethylpropane was used as a substrate to perform the quantum mechanics calculations on LinB (a 33-kDa haloalkane dehalogenase from Sphingomonas paucimobilis UT26) to study which active site conformations and protonation states were eligible to result in catalysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron J Oakley et al.
Biochemistry, 43(4), 870-878 (2004-01-28)
We present the structure of LinB, a 33-kDa haloalkane dehalogenase from Sphingomonas paucimobilis UT26, at 0.95 A resolution. The data have allowed us to directly observe the anisotropic motions of the catalytic residues. In particular, the side-chain of the catalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service