Skip to Content
Merck
All Photos(1)

Key Documents

249637

Sigma-Aldrich

Dimethyl N-cyanodithioiminocarbonate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3S)2C=NCN
CAS Number:
Molecular Weight:
146.23
Beilstein:
635998
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

solid

mp

45-50 °C (lit.)

functional group

amine
thioether

SMILES string

CS\C(SC)=N\C#N

InChI

1S/C4H6N2S2/c1-7-4(8-2)6-3-5/h1-2H3

InChI key

IULFXBLVJIPESI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl N-cyanodithioiminocarbonate has been used in the synthesis of:
  • 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines
  • methylsulfanyl derivatives of azoloazines and azoloazoles
  • methylsulfanylpyrimidines
  • N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles
  • cyanoguanidines

Other Notes

Remainder water

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Mercaptopurine and Thioguanine Analogues: The Reaction of Dimethyl N-Cyanodithioiminocarbonate with Oxo-and Amino-diazoles.
Alqaradawi SY and Elgemeie GH.
Synthetic Communications, 34(5), 805-815 (2004)
A novel synthesis of N-aryl-6-methylsulfanyl-4-pyrimidinones and purine analogues: The reaction of dimethyl N-cyanodithioiminocarbonate with cyanoacetanilides.
Elgemeie GH, et al.
Synthetic Communications, 33(12), 2095-2101 (2003)
First Synthesis of N-Substituted Amino and N-Sulfonylaminated Methylthiopyrimidines: Reaction of Dimethyl N-Cyanodithioiminocarbonate With Substituted Hydrazides.
Elgemeie GH and Sood SA.
Synthetic Communications, 36(6), 743-753 (2006)
The design and synthesis of structurally related mercaptopurine analogues: reaction of dimethyl N-cyano-dithioiminocarbonate with 5-aminopyrazoles.
Elgemeie GH, et al.
Synthetic Communications, 31(22), 3453-3458 (2001)
D Z Hung et al.
Journal of toxicology. Clinical toxicology, 30(3), 351-358 (1992-01-01)
Dimethyl cyanocarbonimidodithioate (CAS No. 10191-60-3) a raw material for cimetidine synthesis, is labelled as an irritant on its storage tank. There is no information available regarding the toxic effects of human exposure. We report a case of severe dermatitis clinically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service