Skip to Content
Merck
All Photos(1)

Documents

142859

Sigma-Aldrich

Difluoroacetic acid

98%

Synonym(s):

1,1-Difluoroacetic acid, 2,2-Difluoroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2CHCO2H
CAS Number:
Molecular Weight:
96.03
Beilstein:
1098588
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.344 (lit.)

bp

132-134 °C (lit.)

mp

−1 °C (lit.)

density

1.526 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C(F)F

InChI

1S/C2H2F2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

PBWZKZYHONABLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Difluoroaceticacid is a monocarboxylic acid and an organofluorine compound. Difluoroaceticacid is easy-to-handle reagent, useful for obtaining difluoromethyl substituents. Difluoroacetic acid can be used asdifluoromethylating reagent for the direct C−H difluoromethylation ofheteroaromatic compounds.

Application

Used in the preparation of 2-difluoromethylbenzimidazoles, -oxazoles and -thiazoles from ortho-substituted anilines mediated by triphenylphoshine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Dalstein et al.
Comptes rendus des seances de la Societe de biologie et de ses filiales, 174(5), 821-825 (1980-01-01)
In the anesthetized rat, the intraperitoneal injection of 40 mg/kg sodium difluoroacetate (DFA), an activator of the pyruvate dehydrogenase, counteracted the hyperlactatemia induced by a high dose of phenformin (40 mg/kg) injected concomitantly. In the normal conscious dog, the administration
Gaëlle Blond et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 8(13), 2917-2922 (2002-12-20)
Ethers of trifluoroacetaldehyde hemiaminals can undergo dehydrofluorination under basic conditions to provide ethers of difluoroketene hemiaminals. The latter behave as equivalents of difluoroacetamide or difluoroacetate anions towards various electrophiles, yielding a range of difluoromethylcarbonyl products.
J N Commandeur et al.
Biochemical pharmacology, 37(23), 4495-4504 (1988-12-01)
The biotransformation and the hepato- and nephrotoxicity of the mercapturic acids (N-acetyl-1-cysteine S-conjugates) of three structurally related 2,2-difluoroethylenes were investigated in vivo in the rat. All mercapturic acids appeared to cause nephrotoxicity, without any measureable effect on the liver. The
Hoffman B M Lantum et al.
Toxicological sciences : an official journal of the Society of Toxicology, 70(2), 261-268 (2002-11-21)
Dichloroacetic acid (DCA), chlorofluoroacetic acid (CFA), and difluoroacetic acid (DFA) are inhibitors of pyruvate dehydrogenase kinase. DCA is used for the clinical management of congenital lactic acidosis. Glutathione transferase zeta (GSTZ1-1) catalyzes the biotransformation of DCA and CFA, and DCA
Tetrahedron Letters, 48, 3251-3251 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service