Skip to Content
Merck
All Photos(1)

Key Documents

245801

Sigma-Aldrich

N-(Hydroxymethyl)acrylamide solution

48 wt. % in H2O

Synonym(s):

Monomethylolacrylamide, N-(Hydroxymethyl)acrylamide, N-Methanolacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONHCH2OH
CAS Number:
Molecular Weight:
101.10
Beilstein:
506646
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

30 ppm monomethyl ether hydroquinone as inhibitor

concentration

48 wt. % in H2O

refractive index

n20/D 1.413

density

1.074 g/mL at 25 °C

SMILES string

OCNC(=O)C=C

InChI

1S/C4H7NO2/c1-2-4(7)5-3-6/h2,6H,1,3H2,(H,5,7)

InChI key

CNCOEDDPFOAUMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-(Hydroxymethyl)acrylamide (NHMA) is a bifunctional monomer with both a reactive vinyl group and a hydroxymethyl group, making it significant in polymer chemistry. It is known for forming hydrophilic, cross-linked networks, with applications in biomedical fields such as drug delivery systems, contact lenses, and tissue engineering, as well as in industrial UV-curable resins. NHMA is commonly used in hydrogel formulations for drug delivery and wound dressings. Additionally, it can be applied in creating bioadhesives or sealants for medical purposes, such as sealing wounds or attaching medical devices to tissues.

Application

N-(Hydroxymethyl)acrylamide can be used as:
  • A monomer in the development of polymer gel dosimeters specifically designed for radiation therapy applications. The incorporation of NHMA enhances the sensitivity of the dosimeter to radiation doses.
  • A monomer in the development of thermo-responsive polymeric gates, which exhibits thermo-responsive behavior. The incorporation of NHMA enhances the overall properties of the polymer, allowing for better control over the release mechanisms in response to temperature changes.
  • A key monomer in the synthesis of double-hydrophilic and amphiphilic block glycopolymers, contributing to their self-assembly properties, biocompatibility, and functionalization potential. These characteristics make NHMA-based glycopolymers valuable for various biomedical applications, including drug delivery systems, biosensors, and tissue engineering.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgit Paulsson et al.
Mutation research, 516(1-2), 101-111 (2002-04-12)
The reactive industrial chemicals acrylamide (AA) and N-methylolacrylamide (MAA) are neurotoxic and carcinogenic in animals, MAA showing a lower potency than AA. The causative agent in AA-induced carcinogenesis is assumed to be the epoxy metabolite, glycidamide (GA), which in contrast
J R Bucher et al.
Journal of toxicology and environmental health, 31(3), 161-177 (1990-11-01)
Toxicology and carcinogenicity studies of N-methylolacrylamide were conducted by administering the chemical by gavage in water to both sexes of F344/N rats and B6C3F1 mice 5 times per week for 16 d, 13 wk, or 2 yr. In 16-d studies
Lars Ole Goffeng et al.
Neurotoxicology and teratology, 30(3), 186-194 (2008-03-21)
The study examines possible persisting effects on the peripheral nervous system and visual system in tunnel workers previously exposed to acrylamide and N-methylolacrylamide during grouting work. We compared neurophysiological function in 44 tunnel workers previously exposed during grouting operations (2-10
Helge Kjuus et al.
Scandinavian journal of work, environment & health, 30(1), 21-29 (2004-03-17)
This study evaluates the possible toxic effects on the peripheral nervous system of tunnel workers exposed to acrylamide and N-methylolacrylamide during grouting work. Symptoms and nerve conduction velocities (NCV) were recorded for 24 tunnel workers 4 and 16 months after
Timothy R Fennell et al.
Toxicological sciences : an official journal of the Society of Toxicology, 71(2), 164-175 (2003-02-04)
Acrylamide (AM) and N-methylolacrylamide (NMA) are used in the formulation of grouting materials. AM undergoes metabolism to a reactive epoxide, glycidamide (GA). Both AM and GA react with hemoglobin to form adducts that can be related to exposure to AM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service