Skip to Content
Merck
All Photos(1)

Key Documents

530042

Sigma-Aldrich

N-Phenylacrylamide

99%

Synonym(s):

Asylanilide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC(O)CH=CH2
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

103-106 °C (lit.)

SMILES string

C=CC(=O)Nc1ccccc1

InChI

1S/C9H9NO/c1-2-9(11)10-8-6-4-3-5-7-8/h2-7H,1H2,(H,10,11)

InChI key

BPCNEKWROYSOLT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Genlin Sun et al.
Journal of separation science, 42(16), 2612-2620 (2019-06-05)
A stationary phase was prepared by chemical derivatization of the support particles with a layer of copolymer composed of styrene and N-phenyl acrylamide. Silica monolith particles of ca. 2.6 µm (volume-based average) have been prepared as the support particles by sol-gel
Pankaj Maharjan et al.
Journal of chromatography. A, 1438, 113-122 (2016-02-26)
Several new types of temperature-responsive ion exchange resins of different polymer composition have been prepared by grafting the products from the co-polymerisation of N-phenylacrylamide, N-iso-propylacrylamide and acrylic acid derivatives onto cross-linked agarose. Analysis of the binding isotherms for these different
Elena V Piletska et al.
Journal of molecular recognition : JMR, 33(4), e2824-e2824 (2019-11-20)
A library of 17 nanoparticles made of acrylate and methacrylate copolymers is prepared, characterized, and screened against six epitopes of adeno-associated viruses (AAV)-neutralizing antibodies to assess their affinity and specificity. Peptide epitopes are immobilized onto the surface of glass beads
Ashraf Ali et al.
Journal of separation science, 40(12), 2654-2661 (2017-05-06)
A specially designed long open tubular capillary column (50 μm internal diameter and 112 cm effective length) was prepared by fabrication of a thin three-component co-polymer layer on the inner surface of silica capillary. A pretreated silica capillary was reacted with 4-(chloromethyl)phenyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service