Skip to Content
Merck
All Photos(2)

Key Documents

235334

Sigma-Aldrich

3,6-Dithia-1,8-octanediol

97%

Synonym(s):

2,2′-(Ethylenedithio)diethanol, Lindlar Catalyst Poison

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)2S(CH2)2S(CH2)2OH
CAS Number:
Molecular Weight:
182.30
Beilstein:
1739193
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

170 °C/0.5 mmHg (lit.)

mp

63-64 °C (lit.)

functional group

hydroxyl
thioether

SMILES string

OCCSCCSCCO

InChI

1S/C6H14O2S2/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

InChI key

PDHFSBXFZGYBIP-UHFFFAOYSA-N

General description

3,6-Dithia-1,8-octanediol is a secondary sulfur-based catalyst poison.

Application

3,6-Dithia-1,8-octanediol has been used:
  • as exogeneous chelator to evaluate a membrane-permeable copper-selective fluorescent sensor for imaging of kinetically labile copper pools
  • in asymmetric total synthesis of (+)-6-epi-castanospermine and polyhydroxylated alkaloid

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liuchun Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(32), 11179-11184 (2005-08-03)
Copper is an essential micronutrient that plays a central role for a broad range of biological processes. Although there is compelling evidence that the intracellular milieu does not contain any free copper ions, the rapid kinetics of copper uptake and
Asymmetric Total Synthesis of (+)-6-epi-Castanospermine by the Stereoselective Formation of a syn, anti Acetylenic 2-Amino-1, 3-diol Stereotriad.
Louvel J, et al.
European Journal of Organic Chemistry, 2010(15), 2921-2926 (2010)
Eric J Stoner et al.
The Journal of organic chemistry, 68(23), 8847-8852 (2003-11-08)
Functionalized erythromycin 9-oxime derivatives are 6-O-allylated under mild conditions using substituted allyl tert-butyl carbonates under palladium(0) catalysis. This allylation works well where traditional ether-forming protocols function poorly. Allyl tert-butyl carbonates provide higher yields in this reaction than lesser substituted carbonates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service