Skip to Content
Merck
All Photos(1)

Key Documents

380474

Sigma-Aldrich

2-Hydroxyethyl disulfide

technical grade

Synonym(s):

2,2′-Dithiodiethanol, Bis(2-hydroxyethyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2SSCH2CH2OH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1735851
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.57 (lit.)

bp

158-163 °C/3.5 mmHg (lit.)

mp

25-27 °C (lit.)

density

1.261 g/mL at 25 °C (lit.)

SMILES string

OCCSSCCO

InChI

1S/C4H10O2S2/c5-1-3-7-8-4-2-6/h5-6H,1-4H2

InChI key

KYNFOMQIXZUKRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxyethyldisulfide (HEDS) is an organosulfur compound ​that belongs to the class ofdialkyl disulfides and has a disulfide bond -S-S- between two hydroxyethylgroups. In material science, HEDS is used as a precursor to form self-assembledmonolayers (SAMs) on gold surfaces for applications such as corrosionprotection, lubrication, and sensors. In addition, 2-hydroxyethyl disulfide isalso used as a reducing agent in organic synthesis and as a crosslinking agentin the production of rubber compounds.

Application

2-Hydroxyethyl disulfide can be used as:
  • A chain extender in the preparation of waterborne self-healing polyurethane, which is used in leather coating.
  • As a precursor in the synthesis of multifunctional initiator, which is used in the preparation of star polystyrenes by atom transfer radical polymerization of styrene.
  • A starting material to prepare a disulfide-based branching agent. This branching agent is used to synthesize highly branched poly(2-hydroxypropyl methacrylate).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Florian Weiland et al.
Frontiers in oncology, 10, 1767-1767 (2020-10-27)
Ovarian cancer is the most lethal gynecologic malignancy. Early detection would improve survival, but an effective diagnostic test does not exist. Novel biomarkers for early ovarian cancer diagnosis are therefore warranted. We performed intraoperative blood sampling from ovarian veins of
Rakesh Kumar et al.
Biomacromolecules, 9(9), 2430-2437 (2008-08-01)
Biodegradable soy protein isolate (SPI), containing 2,2-diphenyl-2-hydroxyethanoic acid, films (SB) were successfully prepared with bis-(2-hydroxyethyl)sulfide as a plasticizer by compression molding at 155 degrees C and 15 MPa. By immersing the SB in distilled water for 26 h, we prepared
P C Hiestand et al.
International journal of immunopharmacology, 7(1), 141-151 (1985-01-01)
ADA 202-718, as well as HEDS and higher homologues of ADA 202-718, were found to profoundly stimulate the delayed type hypersensitivity reaction in mice when given i.p. or orally in a dose range of 0.1 - 10 mg/kg. While even
Nikola Mesecke et al.
Biochemistry, 47(5), 1452-1463 (2008-01-04)
Two novel monothiol glutaredoxins from yeast (ScGrx6 and ScGrx7) were identified and analyzed in vitro. Both proteins are highly suited to study structure-function relationships of glutaredoxin subclasses because they differ from all monothiol glutaredoxins investigated so far and share features
A Goswami et al.
Biochemistry international, 19(2), 361-368 (1989-08-01)
Although all iodothyronine 5'-deiodinases require thiol cofactors for activity, the type II variant has been suspected to contain no reactive thiol groups because of its resistance to inactivation by iodoacetate (IAC). We report here that, under suitable stoichiometric conditions for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service