T30007
2,2′-Thiodiacetic acid
98%
Synonym(s):
2,2′-Thio-bis(acetic acid), Dicarboxydimethyl sulfide, Thiodiglycolic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
S(CH2COOH)2
CAS Number:
Molecular Weight:
150.15
Beilstein:
1764392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
128-131 °C (lit.)
SMILES string
OC(=O)CSCC(O)=O
InChI
1S/C4H6O4S/c5-3(6)1-9-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
InChI key
UVZICZIVKIMRNE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
<ul>
<li><strong>Environmentally Safe Herbicides:</strong> It is used in the synthesis of quaternary ammonium mono- and bis-salts. This compound has been formulated into ammonium 2,2-thiodiacetates, serving as selective and environmentally safe herbicides. Its application underscores its utility in sustainable agriculture and safety in environmental management (Balczewski et al., 2018).</li>
</ul>
<li><strong>Environmentally Safe Herbicides:</strong> It is used in the synthesis of quaternary ammonium mono- and bis-salts. This compound has been formulated into ammonium 2,2-thiodiacetates, serving as selective and environmentally safe herbicides. Its application underscores its utility in sustainable agriculture and safety in environmental management (Balczewski et al., 2018).</li>
</ul>
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
K Norpoth et al.
Journal of cancer research and clinical oncology, 114(2), 158-162 (1988-01-01)
Two groups of male and female Sprague-Dawley rats (50 animals/group per sex) were treated with either 15.37 or 46.77 mumole of 1,1,2-TCE in DMSO/rat for 2 years. The animals were treated once a week by s.c. injection of test compound
Sulekh Chandra et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(8-9), 2153-2162 (2004-07-14)
A novel macrocyclic Schiff base ligand (2,5,9,12,14,18-hexaoxo-7,16-dithia-1,3,4,10,11,13-hexaazacycloocta-decane (H6L) with N4S2 coordinating sites was prepared by the reaction of the semicarbazide and thiodiglycolic acid. The transition metal complexes with macrocyclic ligand were synthesized and characterized by elemental analyses, magnetic susceptibility measurements
Zdenka Dlasková et al.
Analytical and bioanalytical chemistry, 375(1), 164-168 (2003-01-10)
In urine the level of thiodiglycolic acid (TDGA) is a sensitive indicator of exposure of the human organism to some physiologically active or toxic substances, gases and vapors. The determination of TDGA by use of a voltammetric method based on
C Scheick et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 52(1-2), 70-76 (1997-01-01)
Urine samples of premature babies contain high amounts of thiodiacetic acid (TDA). Since these pre-term infants are exposed to an increased oxygen atmosphere in the incubator, we supposed that these high levels of thiodiacetic acid might be produced from ethylene
June Dunnick et al.
Toxicological sciences : an official journal of the Society of Toxicology, 81(1), 243-252 (2004-06-18)
Cardiotoxicity induced by 2-, 3-, 5-, and 12-day dermal administration of 400 and 600 mg/kg/day of bis(2-chloroethoxy)methane to F344/N male and female rats was characterized. The severity and incidence of lesions were similar among males and females and in all
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service