Skip to Content
Merck
All Photos(1)

Key Documents

224715

Sigma-Aldrich

trans-3-Hexen-1-ol

97%

Synonym(s):

trans-3-Hexenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
1719713
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.439 (lit.)

bp

61-62 °C/12 mmHg (lit.)

density

0.817 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

[H]\C(CC)=C(\[H])CCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h3-4,7H,2,5-6H2,1H3/b4-3+

InChI key

UFLHIIWVXFIJGU-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-3-Hexen-1-ol (trans 3-Hexenol) stimulates the antennae of male H. cunea moths[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Tang et al.
PloS one, 7(11), e49256-e49256 (2012-11-21)
Volatile organic compounds (VOCs) were collected from damaged and intact mulberry leaves (Morus alba L., Moraceae) and from Hyphantria cunea larvae by headspace absorption with Super Q columns. We identified their constituents using gas chromatography-mass spectrometry, and evaluated the responses
Shigehiro Namiki et al.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology, 194(5), 501-515 (2008-04-05)
Pheromone-source orientation behavior can be modified by coexisting plant volatiles. Some host plant volatiles enhance the pheromonal responses of olfactory receptor neurons and increase the sensitivity of orientation behavior in the Lepidoptera species. Although many electrophysiological studies have focused on
Andre F Cruz et al.
Phytochemistry, 78, 72-80 (2012-04-24)
Fusarium diseases cause major economic losses in wheat-based crop rotations. Volatile organic compounds (VOC) in wheat and rotation crops, such as chickpea, may negatively impact pathogenic Fusarium. Using the headspace GC-MS method, 16 VOC were found in greenhouse-grown wheat leaves:
Damon J Crook et al.
Journal of economic entomology, 105(2), 429-437 (2012-05-23)
Field trapping assays were conducted in 2009 and 2010 throughout western Michigan, to evaluate lures for adult emerald ash borer, A. planipennis Fairmaire (Coleoptera: Buprestidae). Several ash tree volatiles were tested on purple prism traps in 2009, and a dark
Krista L Ryall et al.
Environmental entomology, 41(3), 648-656 (2012-06-27)
Attraction of emerald ash borer, Agrilus planipennis Fairmaire, to a volatile pheromone was demonstrated in three field experiments using baited green sticky traps. A dose-response curve was generated for male A. planipennis to increasing release rates of (3Z)-dodecen-12-olide ((3Z)-lactone) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service