Skip to Content
Merck
All Photos(2)

Key Documents

188476

Sigma-Aldrich

2-Methylbenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1929783
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.75 mmHg ( 86 °C)

Assay

98%

form

solid

refractive index

n20/D 1.5408 (lit.)

bp

109-110 °C/14 mmHg (lit.)

mp

33-36 °C (lit.)

functional group

hydroxyl

SMILES string

Cc1ccccc1CO

InChI

1S/C8H10O/c1-7-4-2-3-5-8(7)6-9/h2-5,9H,6H2,1H3

InChI key

XPNGNIFUDRPBFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup - (External MSDS)

Flash Point(C)

104 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quinones et al.
Analytical chemistry, 72(7), 1495-1502 (2000-04-14)
High-concentration HPLC band profiles of single solutes and the individual band profiles of the components of binary and ternary mixtures are reported for benzyl alcohol, 2-phenylethanol, and 2-methylbenzyl alcohol. These solutes were eluted from a C18 column by a binary
Non-mutagenicity of toluene, o-, m- and p-xylene, o-methylbenzylalcohol and o-methylbenzylsulfate in the Ames assay.
R P Bos et al.
Mutation research, 88(3), 273-279 (1981-03-01)
Jeong-Eeh Lee et al.
Journal of pharmaceutical and biomedical analysis, 174, 115-122 (2019-06-05)
Benzene (B), toluene (T), ethylbenzene (E), o-, m- and p-xylene (o-, m-, p-X) are ubiquitous and frequently exposed to human throughout the environment. Previously published test methods for phenolic biomarkers are not sensitive enough to be detected in most general

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service