219371
3-Fluorobenzotrifluoride
99%
Synonym(s):
α,α,α,3-Tetrafluorotoluene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
FC6H4CF3
CAS Number:
Molecular Weight:
164.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.4 (lit.)
bp
101-102 °C (lit.)
density
1.302 g/mL at 25 °C (lit.)
functional group
fluoro
SMILES string
Fc1cccc(c1)C(F)(F)F
InChI
1S/C7H4F4/c8-6-3-1-2-5(4-6)7(9,10)11/h1-4H
InChI key
GBOWGKOVMBDPJF-UHFFFAOYSA-N
Application
3-Fluorobenzotrifluoride was used:
- as fluorinated aromatic solvent to investigate electrogenerated chemiluminescence of 9,10-diphenylanthracene, rubrene and anthracene
- used to preparae 2-nitro-5-fluorobenzotrifluoride via nitration reaction
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
44.6 °F - closed cup
Flash Point(C)
7 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aromatic Fluorine Compounds. I. The Synthesis of 2, 5-and 3, 5-Difluorobenzotrifluorides1.
Finger GC and Reed FH.
Journal of the American Chemical Society, 66(11), 1972-1974 (1944)
David J Vinyard et al.
The journal of physical chemistry. A, 112(37), 8529-8533 (2008-08-30)
The electrogenerated chemiluminescence (ECL) of 9,10-diphenylanthracene (DPA), rubrene, and anthracene has been studied in fluorinated aromatic solvents. Mixed annihilation ECL between aromatic luminophores and quinones was observed in solvent systems containing acetonitrile and either benzene, benzotrifluoride, 3-fluorobenzotrifluoride, or 1,3-bis(trifluoromethyl)benzene. Increases
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service