Skip to Content
Merck
All Photos(1)

Documents

15404

Sigma-Aldrich

N-Boc-1,4-butanediamine

≥97.0% (GC/NT)

Synonym(s):

N-Boc-1,4-diaminobutane, tert-Butyl N-(4-aminobutyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONH(CH2)4NH2
CAS Number:
Molecular Weight:
188.27
Beilstein:
1937878
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC/NT)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.460

density

0.984 g/mL at 20 °C (lit.)

functional group

Boc
amine

SMILES string

NCCCCNC(OC(C)(C)C)=O

InChI

1S/C9H20N2O2/c1-9(2,3)13-8(12)11-7-5-4-6-10/h4-7,10H2,1-3H3,(H,11,12)

InChI key

ZFQWJXFJJZUVPI-UHFFFAOYSA-N

Application

  • Carboxy-Silane Coated Iron Oxide Nanoparticles: Details the application of N-Boc-1,4-butanediamine in modifying iron oxide nanoparticles for imaging and drug delivery (D Stanicki, S Boutry, S Laurent, et al., 2014). Access the article.

Other Notes

Preparation of pharmacologically active compounds. Preparation of spermidine analogues. Introduction of a C4-spacer.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Regina Holm et al.
Macromolecular rapid communications, 36(23), 2083-2091 (2015-09-08)
In this work, the synthesis of polypeptoid-block-polypeptide copolymers (block copolypept(o)ides) based on bifunctional initiators is described, which introduces a distinct chemical entity at the connection between both blocks. With a view towards redox-sensitive block copolypept(o)ides, a cystamine-based initiator was used
Hongyan Guo et al.
Journal of medicinal chemistry, 45(10), 2056-2063 (2002-05-03)
Several iron chelators containing alpha,beta-unsaturated hydroxamic acid motifs appended to a citric acid platform were synthesized. Mycobacterium paratuberculosis was then challenged to grow in the presence of a panel of siderophores (mycobactin J, deferrioxamine B, acinetoferrin, and nannochelin A) and
H.H. Wassermann et al
Tetrahedron, 58, 7177-7177 (2002)
Y Shai et al.
Biochemistry, 28(11), 4801-4806 (1989-05-30)
In the present study we synthesize 18F-labeled insulin of high specific radioactivity. A new prosthetic group methodology, in which [18F]fluoride displaces a bromide group of 4-(bromomethyl)-benzoylamine intermediates, was used. The 4-(fluoromethyl)benzoyl product was chemically stable. 18F-Labeled insulin retains the essential
Shaohui Deng et al.
Science advances, 6(29), eabb4005-eabb4005 (2020-08-25)
Controlled release of CRISPR-Cas9 ribonucleoprotein (RNP) and codelivery with other drugs remain a challenge. We demonstrate controlled release of CRISPR-Cas9 RNP and codelivery with antitumor photosensitizer chlorin e6 (Ce6) using near-infrared (NIR)- and reducing agent-responsive nanoparticles in a mouse tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service