Skip to Content
Merck
All Photos(3)

Key Documents

655198

Sigma-Aldrich

Cadmium chloride

technical grade

Synonym(s):

Cadmium dichloride, Dichlorocadmium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

form

solid

concentration

≥55.2% Cd (EDTA titration)

mp

568 °C (lit.)

solubility

H2O: soluble >457 g/L at 20 °C (OECD Test Guideline 105)

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cadmium chloride (CdCl2) may be used as a catalyst for an efficient production of 5-substituted 1H-tetrazoles from [2 + 3]-cycloaddition of NaN3 with nitrile.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cadmium chloride as an efficient catalyst for neat synthesis of 5-substituted 1 H-tetrazoles.
Venkateshwarlu G, et al.
Synthetic Communications, 39(24), 4479-4485 (2004)
Teresa Coccini et al.
Histology and histopathology, 28(2), 195-209 (2013-01-01)
Potential risk associated with new nanomaterial exposure needs to be assessed. This in vivo study investigated pulmonary effects of engineered cadmium-containing silica nanoparticles Cd/SiNPs (1 mg/rat), silica SiNPs (600 μg/rat) and CdCl₂ (400 μg/rat) 1, 7 and 30 days after
Grace Choong et al.
Toxicology and applied pharmacology, 272(2), 423-430 (2013-07-23)
Cadmium disrupts the actin cytoskeleton in rat mesangial cells, and we have previously shown that this involves a complex interplay involving activation of kinase signaling, protein translocation, and disruption of focal adhesions. Here we investigate the role that glutathionylation of
Sujuan Chen et al.
Journal of neurochemistry, 128(2), 256-266 (2013-10-12)
Cadmium (Cd), a toxic environmental contaminant, induces neurodegenerative diseases. Celastrol, a plant-derived triterpene, has shown neuroprotective effects in various disease models. However, little is known regarding the effect of celastrol on Cd-induced neurotoxicity. Here, we show that celastrol protected against
M Auslander et al.
Marine biotechnology (New York, N.Y.), 10(3), 250-261 (2008-01-24)
Individuals of the fish Lithognathus mormyrus were exposed to a series of pollutants including: benzo[a]pyrene, pp-DDE, Aroclor 1254, perfluorooctanoic acid, tributyl-tin chloride, lindane, estradiol, 4-nonylphenol, methyl mercury chloride, and cadmium chloride. Five mixtures of the pollutants were injected. Each mixture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service