Skip to Content
Merck
All Photos(2)

Documents

203718

Sigma-Aldrich

Magnesium oxide

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38110204
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: magnesium

mp

2852 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Eco-friendly synthesis of novel magnesium oxide nanomaterials for dye degradation, battery, and sensor applications: This study explores the synthesis of magnesium oxide nanoparticles and their applications in dye degradation, batteries, and sensors. (Ramakrishna et al., 2024).
  • Impact of silt chemical composition on deflocculation and coagulation of clay-rich paste: The study investigates the role of magnesium oxide in modifying the flocculation properties of clay-rich pastes used in industrial applications. (Ardant et al., 2024).
  • A Systematic Review of Synthesis MgO Nanoparticles and Their Applications: This review covers various methods for the synthesis of magnesium oxide nanoparticles and their diverse applications. (Hemmami et al., 2024).
  • Comparative investigation on physicochemical, biological, and cytotoxicity of magnesium oxide prepared by chemically mediated and green synthesis: This research compares different synthesis methods of magnesium oxide nanoparticles and their effects on physicochemical properties, biological activity, and cytotoxicity. (Archana & Menon, 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from
Xiaoyan Sun et al.
BMC plant biology, 15, 116-116 (2015-05-11)
Heat stress is a critical threat to tall fescue in transitional and warm climate zones. Identification of association between molecular markers and heat tolerance-related functional traits would promote the efficient selection of heat tolerant tall fescue cultivars. Association analysis of
Sanket A Deshmukh et al.
Physical chemistry chemical physics : PCCP, 14(44), 15593-15605 (2012-10-19)
Atomic scale characterization of the structure and dynamics of confined water molecules located near the metal oxide-aqueous interface is carried out using molecular dynamics simulations. Proximity effects on water molecules (H(2)O) near a magnesium oxide surface (MgO(100)) at room temperature
Jian-Bin Shi et al.
Carbohydrate research, 359, 65-69 (2012-08-29)
This work describes the structural changes of bagasse hemicelluloses during the cooking process involving active oxygen (O(2) and H(2)O(2)) and solid alkali (MgO). The hemicelluloses obtained from the bagasse raw material, pulp, and yellow liquor were analyzed by high-performance anion-exchange
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service