Skip to Content
Merck
All Photos(2)

Documents

529699

Sigma-Aldrich

Magnesium oxide

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

powder

mp

2852 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium oxide (MgO) is a metal oxide formed by processing magnesite. It can also be extracted from sea water. It is industrially produced by the calcination of magnesium carbonate or magnesium hydroxide.

Application

MgO metal powder can be used for a variety of applications such as medical use, refractory materials, energy applications and in the preparation of magnesia nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Magnesium oxide/water chemical heat pump to enhance energy utilization of a cogeneration system
Kato Y, et al.
Energy, 30(11-12), 2144-2155 (2005)
Hydration of refractory oxides in castable bond systems?I: alumina, magnesia, and alumina-magnesia mixtures
Ahari KG, et al.
J. Eur. Ceram. Soc., 22(4), 495-503 (2002)
The Chemistry and Technology of Magnesia, 210 (2006)
Medical treatments for the maintenance therapy of reflux oesophagitis and endoscopic negative reflux disease
Donnellan C, et al.
Cochrane database of systematic reviews (Online), 210(4) (2004)
Magnesium oxide nanoparticles prepared by ultrasound enhanced hydrolysis of Mg-alkoxides
Stengl V, et al.
Materials Letters, 57(24-25), 3998-4003 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service