Merck
  • The structural changes of the bagasse hemicelluloses during the cooking process involving active oxygen and solid alkali.
The structural changes of the bagasse hemicelluloses during the cooking process involving active oxygen and solid alkali.
Carbohydrate research (2012-08-29)
Jian-Bin Shi, Qiu-Lin Yang, Lu Lin, Jun-Ping Zhuang, Chun-Sheng Pang, Tu-Jun Xie, Ying Liu
ABSTRACT
This work describes the structural changes of bagasse hemicelluloses during the cooking process involving active oxygen (O(2) and H(2)O(2)) and solid alkali (MgO). The hemicelluloses obtained from the bagasse raw material, pulp, and yellow liquor were analyzed by high-performance anion-exchange chromatography (HPAEC), gel permeation chromatography (GPC), Fourier transform infrared spectroscopy (FT-IR), and (1)H-(13)C 2D hetero-nuclear single quantum coherence spectroscopy (HSQC). The results revealed that the structure of the bagasse hemicelluloses was L-arabino-(4-O-methylglucurono)-D-xylan. Some sugar units in hemicelluloses were oxidized under the cooking conditions. Additionally, the backbones and the ester linkages of hemicelluloses were heavily cleaved during the cooking process.
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh