Skip to Content
Merck
All Photos(3)

Key Documents

202711

Sigma-Aldrich

Barium carbonate

99.999% trace metals basis

Synonym(s):

Barium monocarbonate, Witherite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCO3
CAS Number:
Molecular Weight:
197.34
Beilstein:
7045119
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38020211
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

[Ba++].[O-]C([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium carbonate is an inorganic compound that is poorly soluble in water. It is used in various industrial applications, particularly in the ceramics industry, where it is used in glazes and as a fluxing agent. It is also used in the manufacture of electronic ceramics, capacitors, PTC thermistors, and other electronic components.

Application

Barium carbonate can be used:    
  • A precursor for the preparation of BaTiO3 nanoparticles, a dielectric material which is useful for manufacturing multilayer ceramic capacitors.
  • A precursor for the synthesis of barium titanate thin films, which finds applications in photovoltaic devices.

Barium carbonate can also be used in following applications:
  • BaCO3 nanoparticles have been reported as catalysts for high-temperature oxygen reduction reactions (ORR) in solid oxide fuel cells (SOFCs).      
  • Barium carbonate-based nanomaterials have been employed for sensor and catalysis applications, with improved sensing performance using one-dimensional nanostructures.    
  • BaCO3 is used to make barium titanate for Multi-layer ceramic capacitor and composite oxides, as well as for PTC thermistors. It′s also used to make optical glass, sputter glass for semiconductors, and as a component of phosphor materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A modified method for barium titanate nanoparticles synthesis
Ashiri R, et al.
Mat. Res. Bul., 46, 2291?2295-2291?2295 (2011)
Miyako Akiyoshi et al.
Journal of hazardous materials, 132(2-3), 141-147 (2005-12-14)
Iron picrate (FePic) was synthesized under conditions similar to those that result in the natural deterioration of chemical weapons. Its thermal hazard was investigated by comparing it with iron picrate obtained by the chemical synthesis method (FePic(Ba)). FePic has eight
Wenjie Zhu et al.
Chemical communications (Cambridge, England), 48(68), 8544-8546 (2012-07-19)
We discovered that micelles of a thermo-responsive polypeptide-based copolymer are able to direct growth of barium carbonate (BaCO(3)) in the form of nanobelts. The BaCO(3) nanobelts tend to grow around a formed crystal, and curl into a spiral superstructure.
Akemi Yasukawa et al.
Journal of colloid and interface science, 288(2), 468-474 (2005-06-02)
Particles of carbonated barium-calcium hydroxyapatite solid solutions (BaCaHap) with different Ba/(Ba+Ca) (X(Ba)) atomic ratios were prepared by a wet method at 100 degrees C and characterized by various means. The crystal phases and structures of the products strongly depended on
Shuisheng Wu et al.
Inorganic chemistry, 48(21), 10326-10329 (2009-09-29)
In this paper, we report on biomineralization of BaCO(3) hierarchical architectures with self-cleaning ability. The phase structures of the obtained samples were characterized by X-ray diffraction (XRD). All of these complex nanostructures, including dendrite-like nanostructures, dumbbell-like nanostructures, and spherical nanostructures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service