Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead(II) oxide
Synonym(s):
Lead(II) oxide
Linear Formula:
PbO
CAS Number:
1317-36-8
Molecular Weight:
223.20
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (4)
Supelco (2)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
211907
Lead(II) oxide,
powder, <10 μm,
ReagentPlus
®
, ≥99.9% trace metals basis
Expand
View Pricing
402982
Lead(II) oxide,
ACS reagent, ≥99.0%
Expand
View Pricing
203610
Lead(II) oxide,
99.999% trace metals basis
Expand
View Pricing
15338
Lead(II) oxide,
puriss. p.a., ≥99.0% (KT), yellow
Expand
View Pricing
1.07401
Lead(II) oxide,
for analysis EMSURE
®
Expand
View Pricing
1.05658
Lead(II) oxide,
EMPLURA
®
Expand
View Pricing