Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Barium carbonate
Synonym(s):
Barium carbonate
Linear Formula:
BaCO
3
CAS Number:
513-77-9
Molecular Weight:
197.34
Products
Product Category
inorganic bases (3)
salts (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
237108
Barium carbonate,
ACS reagent, ≥99%
Expand
View Pricing
202711
Barium carbonate,
99.999% trace metals basis
Expand
View Pricing
329436
Barium carbonate,
99.98% trace metals basis
Expand
View Pricing
1.01714
Barium carbonate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1.01711
Barium carbonate,
Technipur
®
Expand
View Pricing