Skip to Content
Merck
All Photos(1)

Key Documents

184268

Sigma-Aldrich

4-tert-Butylbenzyl alcohol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CH2OH
CAS Number:
Molecular Weight:
164.24
Beilstein:
1860665
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.517 (lit.)

bp

140 °C/20 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)c1ccc(CO)cc1

InChI

1S/C11H16O/c1-11(2,3)10-6-4-9(8-12)5-7-10/h4-7,12H,8H2,1-3H3

InChI key

FVEINXLJOJPHLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylbenzyl alcohol forms inclusion complex with β-cyclodextrin and crystal structure of complex has been studied. Selective oxidation of 4-tert-butylbenzyl alcohol by hypochlorite and phase transfer catalyst has been investigated in a membrane reactor.

Application

4-tert-Butylbenzyl alcohol was used in preparation of novel copolyimides bearing bulky pendent groups by chemical modification of a copolyimide precursor containing carboxylic acid groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

262.4 °F - DIN 51758

Flash Point(C)

128 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of the inclusion complex of cyclomaltoheptaose (?-cyclodextrin) with 4-tert-butylbenzoic acid.
Rontoyianni A, et al.
Carbohydrate Research, 252, 19-32 (1994)
The selective oxidation of benzyl alcohols in a membrane reactor.
Grigoropoulou G, et al.
Chemical Communications (Cambridge, England), 6, 547-548 (2001)
Chemical modification of copolyimides with bulky pendent groups: effect of modification on solubility and thermal stability.
Maya EM, et al.
Polymer Degradation and Stability, 92(12), 2294-2299 (2007)
Tsunehisa Hirose et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-05)
The retention behavior of a wide variety of stationary phases was compared in supercritical fluid chromatography (SFC) and normal-phase high-performance liquid chromatography (NP-HPLC). We also attempted to elucidate the retention behavior in SFC by investigating the selectivity of the different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service