Skip to Content
Merck
All Photos(3)

Key Documents

P13800

Sigma-Aldrich

1-Phenylethanol

98%

Synonym(s):

(±)-1-Phenylethanol, (±)-α-Methylbenzyl alcohol, Methyl phenyl carbinol, Styrallyl alcohol, Styrene alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH3
CAS Number:
Molecular Weight:
122.16
Beilstein:
1905149
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.21 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

98%

refractive index

n20/D 1.527 (lit.)

bp

204 °C/745 mmHg (lit.)

mp

19-20 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

CC(O)c1ccccc1

InChI

1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3

InChI key

WAPNOHKVXSQRPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 1-Phenylethanol can be used in the acid catalyzed Ritter reaction to synthesize various N-benzylacetamides.
  • It is one of the key precursors for the synthesis of [60] fullerene-fused isochroman derivatives via palladium-catalyzed hydroxyl-directed cyclization.
  • It is used in Friedlander synthesis of quinolines where 1-phenylethanol reacts with 2-aminobenzylic alcohol derivatives to yield corresponding quinones.
  • It can also be used in the regioselective, iridium-catalyzed multicomponent synthesis of pyrimidines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

187.9 °F - Pensky-Martens closed cup

Flash Point(C)

86.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of [60] fullerene-fused tetrahydrobenzooxepine and isochroman derivatives via hydroxyl-directed C?H activation/C?O cyclization.
Zhai W Q, et al.
Organic Letters, 16(6), 1638-1641 (2014)
Transition-metal-free indirect friedlander synthesis of quinolines from alcohols.
Martinez R, et al.
The Journal of Organic Chemistry, 73(24), 9778-9780 (2008)
Pedro Lozano et al.
Biotechnology letters, 28(19), 1559-1565 (2006-08-11)
Continuous dynamic kinetic resolution processes in different ionic liquid/supercritical carbon dioxide biphasic systems were carried out by simultaneously using both immobilized Candida antarctica lipase B (Novozym 435) and silica modified with benzenosulfonic acid (SCX) catalysts at 40 degrees C and
The Ritter reaction under truly catalytic Br?nsted acid conditions.
Sanz R, et al.
European Journal of Organic Chemistry, 2007(28), 4642-4645 (2007)
A sustainable multicomponent pyrimidine synthesis.
Deibl N, et al.
Journal of the American Chemical Society, 137(40), 12804-12807 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service