Skip to Content
Merck
All Photos(1)

Key Documents

186015

Sigma-Aldrich

Bicyclo[3.3.1]nonan-9-one

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

solid

mp

155-157 °C (lit.)

functional group

ketone

SMILES string

O=C1[C@@H]2CCC[C@H]1CCC2

InChI

1S/C9H14O/c10-9-7-3-1-4-8(9)6-2-5-7/h7-8H,1-6H2/t7-,8+

InChI key

SKTMMSQPXGWCAP-OCAPTIKFSA-N

Related Categories

General description

Carbon-13 NMR spectra of solid polycrystalline bicyclo[3.3.1]nonan-9-one has been studied at 315K.

Application

Bicyclo[3.3.1]nonan-9-one was used in the preparation of bicyclo[3.3.1]nonan-2-ene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon-13 NMR spectra of solid bicyclo [3. 3. 1] nonan-9-one. Conformational studies in the solid state.
Wasylishen RE and Friesen KJ.
Org. Magn. Reson., 13(5), 343-344 (1980)
Proximity Effects. XLIII. The Solvolysis of 4-Cyclooctene-1-methyl Brosylate1, 2
Cope AC, et al.
Journal of the American Chemical Society, 87(14), 3130-3135 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service