Skip to Content
Merck
All Photos(3)

Key Documents

146048

Sigma-Aldrich

2-Adamantanone

ReagentPlus®, 99%

Synonym(s):

2-Oxoadamantane, Tricyclo[3.3.1.13,7]decanone (9CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
1210235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

product line

ReagentPlus®

Assay

99%

form

solid

mp

256-258 °C (subl.) (lit.)

functional group

ketone

SMILES string

O=C1C2CC3CC(C2)CC1C3

InChI

1S/C10H14O/c11-10-8-2-6-1-7(4-8)5-9(10)3-6/h6-9H,1-5H2/t6-,7+,8-,9+

InChI key

IYKFYARMMIESOX-SPJNRGJMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Adamantanone on deprotonation in the gas phase affords the corresponding β-enolate anion[1]. It reacts with 1,1-dilithio-1-sila-2,3,4,5-tetraphenylsilole to yield 5-silafulvene[2].

Application

2-Adamantanone was used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane[3] and (+/-)-1-(adamantan-2-yl)-2-propanamine[4].

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew M Meyer et al.
The Journal of organic chemistry, 75(12), 4274-4279 (2010-05-26)
Deprotonation of 2-adamantanone (1) in the gas phase affords the corresponding beta-enolate anion. This ion was independently prepared by the fluoride-induced desilylation of 4-trimethylsilyl-2-adamantanone, and its reactivity and thermodynamic properties were measured (DeltaH degrees(acid) = 394.7 +/- 1.4, EA =
Sunil Sabbani et al.
Bioorganic & medicinal chemistry letters, 18(21), 5804-5808 (2008-10-11)
Dispiro N-Boc-protected 1,2,4-trioxane 2 was synthesised via Mo(acac)(2) catalysed perhydrolysis of N-Boc spirooxirane followed by condensation of the resulting beta-hydroperoxy alcohol 10 with 2-adamantanone. N-Boc 1,2,4-trioxane 2 was converted to the amine 1,2,4-trioxane hydrochloride salt 3 which was subsequently used
Yulan Chen et al.
Nature chemistry, 4(7), 559-562 (2012-06-22)
Nature uses mechanochemical transduction processes to achieve diverse and vital functions, such as hearing, cellular adhesion and gating of ion channels. One fascinating example of biological mechanotransduction is the emission of light on mechanical stimulation. However, molecular-level transduction of force
Hassen Jaafar et al.
Dalton transactions (Cambridge, England : 2003), 40(1), 92-106 (2010-11-19)
We report that the oxygen sensitivity of some Fe(II) complexes with tripodal ligands can be used, with benefit, in the oxidation of cyclohexane under mild conditions. Depending on the solvent, two very different reaction pathways are involved, which share the
Irina S Toulokhonova et al.
Journal of the American Chemical Society, 126(17), 5336-5337 (2004-04-29)
Reaction of 1,1-dilithio-1-sila-2,3,4,5-tetraphenylsilole with 2-adamantanone produces a 5-silafulvene. This represents a new method for synthesis of silenes, leading to the first example of a silapentafulvene.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service