Skip to Content
Merck
All Photos(1)

Key Documents

T19208

Sigma-Aldrich

β-Tetralone

98%

Synonym(s):

beta-Tetralone, 3,4-Dihydro-2(1H)-naphthalenone, 3,4-Dihydro-2-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
Beilstein:
509386
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.560 (lit.)

bp

131 °C/11 mmHg (lit.)

mp

18 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CCc2ccccc2C1

InChI

1S/C10H10O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-4H,5-7H2

InChI key

KCKZIWSINLBROE-UHFFFAOYSA-N

Gene Information

human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed asymmetric allylic alkylation of alpha-aryl ketones.
Barry M Trost et al.
Angewandte Chemie (International ed. in English), 41(18), 3492-3495 (2002-09-26)
H S Shin et al.
Applied microbiology and biotechnology, 57(4), 506-510 (2002-01-05)
Biotransformation of 6-bromo-2-tetralone (Br-beta-tetralone) to 6-bromo-2-tetralol (Br-beta-tetralol) by yeast cells of Trichosporon capitatum (ATCC 74312) and its partially purified Br-beta-tetralone reductase was evaluated in an electrochemical bioreactor. The biotransformation rates and final product formation were significantly affected by substrate concentration
R D Mattes et al.
Physiology & behavior, 72(1-2), 217-229 (2001-03-10)
Ethanol is a common dietary constituent, but knowledge of its chemosensory properties and their relationship to its ingestion is limited. Twenty-five male and 25 female, light-regular ethanol consumers participated in five test sessions. Sessions involved determination of taste, olfactory and
M Goto et al.
Chemical & pharmaceutical bulletin, 48(10), 1529-1531 (2000-10-25)
The reactions of the lithium enolate of 1-tetralone with reactive alkyl halides were examined in the absence and in the presence of 3 eq of various ligands for the lithium. It is shown that the rates of the reactions are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service