Skip to Content
Merck
All Photos(3)

Key Documents

185426

Sigma-Aldrich

o-Toluidine

≥99%

Synonym(s):

2-Aminotoluene, 2-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
741981
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

0.26 mmHg ( 25 °C)

Assay

≥99%

form

liquid

autoignition temp.

899 °F

refractive index

n20/D 1.572 (lit.)

bp

199-200 °C (lit.)
89-90 °C/11 mmHg (lit.)

mp

−28 °C (lit.)

solubility

alcohol: soluble
diethyl ether: soluble
water: slightly soluble

density

1.008 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1N

InChI

1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3

InChI key

RNVCVTLRINQCPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

o-Toluidine undergoes chemically oxidative polymerization with aniline in the presence of ammonium persulfate as an oxidant in HCl medium.

Application

o-Toluidine was used in determination of glucose in biological materials. It was used as precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A T Smit et al.
Bioresource technology, 243, 994-999 (2017-07-30)
Enzymatic cellulose hydrolysis of pretreated wheat straw pulp to glucose is enhanced when the hydrolysis is performed in the presence of an aqueous extract of the wheat straw. A relative digestibility increase of about 10% has been observed for organosolv
In situ synthesis of copper nanoparticles and poly (< i> o</i>-toluidine): A metal-polymer composite material.
Mallick K, et al.
Eur. Polymer J., 42(3), 670-675 (2006)
Oxidative copolymers of aniline with o-toluidine: Their structure and thermal properties.
Huang M-R, et al.
Journal of Applied Polymer Science, 81(8), 1838-1847 (2001)
Ivo Kalkman et al.
Physical chemistry chemical physics : PCCP, 11(21), 4311-4318 (2009-05-22)
The rotationally resolved spectrum of the o-toluidine S(1)<-- S(0) origin was measured using laser induced fluorescence spectroscopy. From the resulting spectrum torsional barriers to internal rotation of the methyl group were derived, which resulted in S(0) state values of V(3)
Emna Hmani et al.
Journal of hazardous materials, 170(2-3), 928-933 (2009-06-23)
Electrochemical oxidation of O-toluidine (OT) was studied by galvanostatic electrolysis using lead dioxide (PbO2) and boron-doped diamond (BDD) as anodes. The influence of operating parameters, such as current density, initial concentration of OT and temperature was investigated. Measurements of chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service