I12808
Isatoic anhydride
96%
Synonym(s):
3,1-Benzoxazine-2,4(1H)-dione, Anthranilic acid N-carboxylic acid anhydride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C8H5NO3
CAS Number:
Molecular Weight:
163.13
Beilstein:
136786
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor density
5.6 (vs air)
Assay
96%
mp
233 °C (dec.) (lit.)
SMILES string
O=C1Nc2ccccc2C(=O)O1
InChI
1S/C8H5NO3/c10-7-5-3-1-2-4-6(5)9-8(11)12-7/h1-4H,(H,9,11)
InChI key
TXJUTRJFNRYTHH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
586.4 °F - closed cup
Flash Point(C)
308 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J E Churchich
Analytical biochemistry, 213(2), 229-233 (1993-09-01)
Isatoic anhydride reacts with nucleophile groups of proteins to yield o-aminobenzoyl protein conjugates. The fluorescence emitted by the chromophore decays in a multiexponential manner with average fluorescence lifetimes ranging from 9.9 to 10.7 ns. The steady emission anisotropy, measured upon
Zhan-Hui Zhang et al.
Journal of combinatorial chemistry, 12(5), 643-646 (2010-08-06)
A simple and efficient protocol for one-pot three-component coupling of isatoic anhydride, amines, and aldehydes in water using magnetically recoverable Fe(3)O(4) nanoparticles is reported. This methodology results in the synthesis of a variety of 2,3-dihydroquinazolin-4(1H)-ones in high yields. The catalyst
M G Page
The Biochemical journal, 295 ( Pt 1), 295-304 (1993-10-01)
Class C beta-lactamases from Pseudomonas aeruginosa and several species of the Enterobacteriaceae have been observed to undergo a rapid burst in hydrolysis of beta-lactam antibiotics before relaxation to a steady-state rate of hydrolysis. The amplitude of the burst corresponds to
Suicide enzyme inactivators.
R H Abeles
Basic life sciences, 25, 287-305 (1983-01-01)
Christine Hiu-Tung Chen et al.
Molecular diversity, 9(4), 353-359 (2005-11-29)
Reactions using fluorous reagents and scavengers are compared side-by-side with their solid-supported counterparts. Fluorous triphenylphosphine is used in the bromination reaction of alcohols, fluorous thiol is used as an electrophile scavenger for alpha-bromoketones, fluorous isatoic anhydride is used as a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service