Skip to Content
Merck
All Photos(2)

Key Documents

183938

Sigma-Aldrich

2,3,5,6-Tetramethylpyrazine

98%

Synonym(s):

2,3,5,6-Tetramethylpyrazine, Ligustrazin, Ligustrazine, Tetrapyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
Beilstein:
113100
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39161809
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

190 °C (lit.)

mp

77-80 °C (lit.)

SMILES string

Cc1nc(C)c(C)nc1C

InChI

1S/C8H12N2/c1-5-6(2)10-8(4)7(3)9-5/h1-4H3

InChI key

FINHMKGKINIASC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5,6-Tetramethylpyrazine has been isolated from Ephedra sinica. It is useful in the treatment of several disorders such as asthma, heart failure, rhinitis and urinary incontinence. It is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gyoseon Goya M Yeom et al.
International immunopharmacology, 18(2), 262-269 (2013-12-18)
2,3,5,6-Tetramethylpyrazine (TMP) is known as a composition of Ephedra sinica and it has been used in the treatment of several disorders such as asthma, heart failure, rhinitis, and urinary incontinence. It has been reported that TMP inhibits melanoma metastasis and
Christopher R E Coggins et al.
Inhalation toxicology, 23 Suppl 1, 84-89 (2011-04-05)
Three heterocyclic nitrogen compounds, 2,3-diethylpyrazine (DEP), 2,3,5,6-tetramethylpyrazine (TMP), and 2-acetyl pyridine (AP), are naturally present in tobacco and are also added to tobacco as flavor ingredients. A battery of tests was used to compare the toxicity of mainstream smoke from
B R Sreekanth et al.
Chemical communications (Cambridge, England), (23)(23), 2375-2377 (2007-09-12)
Co-crystals of 4-hydroxybenzoic acid and 2,3,5,6-tetramethylpyrazine (2 : 1) exhibit the first supramolecular synthon polymorphism in a co-crystal; metastable anti-hierarchic polymorph I converts to stable hierarchic form II.
Xian-Chao Cheng et al.
Bioorganic & medicinal chemistry, 17(8), 3018-3024 (2009-03-31)
A series of novel acylpiperazinyl Ligustrazine derivatives was designed, synthesized, and their protective effects on damaged ECV-304 cells and antiplatelet aggregation activities were evaluated. The results showed that compound E33 displayed most potential protective effects on the ECV-304 cells damaged
Huda M AlKreathy et al.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society, 28(8), 916-926 (2020-08-15)
Available antiulcer medications reveal partial efficacy and numerous adverse reactions. Tetramethylpyrazine (TMP) was known for its potential antioxidant, anti-inflammatory and angiogenic properties. The aim of current study was to investigate the potential gastroprotective effect of TMP against indomethacin-induced gastric ulcer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service