Skip to Content
Merck
All Photos(1)

Key Documents

G8503

Sigma-Aldrich

18α-Glycyrrhetinic acid

≥95%

Synonym(s):

18α-Glycyrrhetic acid, 18-Isoglycyrrhetinic acid, 3β-Hydroxy-11-oxo-18α,20β-olean-12-en-29-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46O4
CAS Number:
Molecular Weight:
470.68
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

SMILES string

[H][C@]12C[C@](C)(CC[C@]1(C)CC[C@]3(C)C2=CC(=O)C4[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C)C(O)=O

InChI

1S/C30H46O4/c1-25(2)21-8-11-30(7)23(28(21,5)10-9-22(25)32)20(31)16-18-19-17-27(4,24(33)34)13-12-26(19,3)14-15-29(18,30)6/h16,19,21-23,32H,8-15,17H2,1-7H3,(H,33,34)/t19-,21+,22+,23-,26-,27+,28+,29-,30-/m1/s1

InChI key

MPDGHEJMBKOTSU-PMTKVOBESA-N

Looking for similar products? Visit Product Comparison Guide

General description

18α-Glycyrrhetinic acid (18α-GA) is a bioactive triterpenoid found in licorice. It shows selective inhibition of 11-HSD1 (11-hydroxysteroid dehydrogenase 1). It also shows anti-proliferative and apoptotic effect on hepatic stellate cells (HSCs).

Application

18α-Glycyrrhetinic acid may be used to synthesize:
  • 3-keto-18α-glycyrrhetinic acid
  • methyl esters of 18α-glycyrrhetinic acid
  • methyl esters of 3-keto-18α-glycyrrhetinic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian M Goerke et al.
Tissue engineering. Part A, 18(23-24), 2395-2405 (2012-06-27)
Neovascularization represents an important issue in tissue-engineering applications, since survival of implanted cells strongly relies on sufficient oxygen and nutrient supply. We have recently observed that human bone marrow-derived mesenchymal stem cells (MSCs) support neovessel formation originating from coimplanted endothelial
Selective inhibition of 11?-hydroxysteroid dehydrogenase 1 by 18a-glycyrrhetinic acid but not 18?-glycyrrhetinic acid.
Classen-Houben D, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 113(3), 248-252 (2009)
Manuela Buonanno et al.
PloS one, 6(6), e21540-e21540 (2011-07-09)
An increased risk of carcinogenesis caused by exposure to space radiation during prolonged space travel is a limiting factor for human space exploration. Typically, astronauts are exposed to low fluences of ionizing particles that target only a few cells in
18a-glycyrrhetinic acid extracted from Glycyrrhiza radix inhibits proliferation and promotes apoptosis of the hepatic stellate cell line.
Zong L, et al.
Journal of Digestive Diseases, 14(6), 328-336 (2013)
Simultaneous quantification of flavonoids and triterpenoids in licorice using HPLC.
Wang YC and Yang YS.
Journal of Chromatography. B, Biomedical Applications, 850(1), 392-399 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service