Skip to Content
Merck
All Photos(1)

Documents

156930

Sigma-Aldrich

Lauroyl chloride

98%

Synonym(s):

Dodecanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COCl
CAS Number:
Molecular Weight:
218.76
Beilstein:
1281201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

134-137 °C/11 mmHg (lit.)

solubility

methanol: soluble 100 mg/mL, clear

density

0.946 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC(Cl)=O

InChI

1S/C12H23ClO/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3

InChI key

NQGIJDNPUZEBRU-UHFFFAOYSA-N

Application

Lauroyl chloride was used:
  • as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals
  • in the preparation of acylated collagen with water solubility and better surface activity
  • as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel
  • in the synthesis of hemicellulose-based hydrophobic biomaterials

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parvaiz Ahmad Bhat et al.
Food chemistry, 340, 128168-128168 (2020-10-05)
The chemical instability of Citral in acidic conditions is viewed as hurdle to commercialize it in food/beverage industries. We attempted to stabilize citral in various single and mixed surfactant systems at pH 1.0 and temperature 25 °C. The study highlights the
Ana G Cunha et al.
Langmuir : the ACS journal of surfaces and colloids, 30(31), 9327-9335 (2014-07-22)
Nanocelluloses are bio-based nanoparticles of interest as stabilizers for oil-in-water (o/w) Pickering emulsions. In this work, the surface chemistry of nanocelluloses of different length, nanofibrillated cellulose (NFC, long) and cellulose nanocrystals (CNC, short), was successfully tailored by chemical modification with
Feng Peng et al.
Carbohydrate research, 343(17), 2956-2962 (2008-09-17)
Hemicellulose-based hydrophobic biomaterials with degrees of substitution ranging from 0.46 to 1.54 were synthesized under mild conditions in homogeneous media (N,N-dimethylformamide-lithium chloride) by reacting the native wheat straw hemicellulosic polymers with lauroyl chloride using 4-dimethylaminopyridine as a catalyst. Other catalysts
Conghu Li et al.
International journal of biological macromolecules, 57, 92-98 (2013-03-19)
A new kind of acylated collagen with water solubility and better surface activity was prepared via reaction of pepsin-solubilized calf skin collagen with lauroyl chloride and succinic anhydride in this paper. The equilibrium surface tension and the isoelectric point were
Michelle M Blum et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 100(7), 1755-1763 (2012-07-19)
A novel material design was developed by functionalizing polyvinyl alcohol hydrogel with an organic low-friction boundary lubricant (molar ratios of 0.2, 0.5, and 1.0 moles of lauroyl chloride). The hydrogels were fabricated using two different techniques. First, the boundary lubricant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service