171158
Stearoyl chloride
97%
Synonym(s):
Octadecanoyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3(CH2)16COCl
CAS Number:
Molecular Weight:
302.92
Beilstein:
639784
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050509
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.454 (lit.)
bp
174-178 °C/2 mmHg (lit.)
mp
21-22 °C (lit.)
density
0.897 g/mL at 25 °C (lit.)
functional group
acyl chloride
SMILES string
CCCCCCCCCCCCCCCCCC(Cl)=O
InChI
1S/C18H35ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3
InChI key
WTBAHSZERDXKKZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Stearoyl chloride was used in the synthesis of 4-fluoroceramide. It was also used in the preparation of shimofuridin analogs: 2′-O-(4-O-stearoyl-alpha-L-fucopyranosyl)thymidine and -uridine.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jun Ning et al.
Carbohydrate research, 338(1), 55-60 (2002-12-31)
Two shimofuridin analogs: 2'-O-(4-O-stearoyl-alpha-L-fucopyranosyl)thymidine (2) and -uridine (3) have been synthesized using D-arabinose, L-fucose, thymine, uracil, and stearoyl chloride as the starting materials. The synthetic procedures involve the facile preparation of 1-(3,5-di-O-benzyl-beta-D-ribofuranosyl)thymine (9) and -uracil (10) by coupling of 1,2-anhydro-3,5-di-O-benzyl-alpha-D-ribofuranose
Gergana S Nikolova et al.
Beilstein journal of organic chemistry, 4, 12-12 (2008-10-23)
Sphingolipids belong to the most important constituents of the membranes of eukaryotic cells. As intermediates in sphingolipid metabolism, sphingosine and its N-octadecanoyl-derivative, ceramide, exhibit a variety of biological functions. These compounds play a crucial role in many essential biological processes
A M Ladhoff et al.
Biomedica biochimica acta, 43(7), 963-969 (1984-01-01)
The assembly of electron dense ferritin modified by acylation with steaorylchloride into small and large egg lecithin vesicles is reported. From electron micrographs conclusions are drawn: on the mode of ferritin incorporation in the lipid bilayer: Small liposomes seem to
[Chemical modification of proteins (enzymes) by water-insoluble reagents].
A V Levashov et al.
Doklady Akademii nauk SSSR, 278(1), 246-248 (1984-09-01)
A V Kabanov et al.
Biomedical science, 1(1), 63-67 (1990-01-01)
A method is proposed for the inhibition of viral reproduction in cells by means of fatty-acylated antiviral antibodies which, in contrast to the unmodified antibodies, have the ability to enter the cells. The potential of this technique is demonstrated in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service