Skip to Content
Merck
All Photos(1)

Documents

M2209

Sigma-Aldrich

(R)-(−)-Mandelic acid

98%

Synonym(s):

(R)-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2691094
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]25/D −151°, c = 1 in ethanol

optical purity

ee: 99% (GLC)

mp

131-133 °C (lit.)

SMILES string

O[C@@H](C(O)=O)c1ccccc1

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1

InChI key

IWYDHOAUDWTVEP-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>374.0 °F

Flash Point(C)

> 190 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Monier et al.
International journal of biological macromolecules, 55, 207-213 (2013-01-30)
An enantioselective S-mandelic acid (S-MA) imprinted chitosan (SMIC) was prepared by cross-linking of chitosan using formaldehyde cross-linker, in the presence of S-MA as an imprint template molecule and 0.5% acetic acid solution as a solvent. Non-imprinted cross-linked chitosan (NIC) as
P G Jaison et al.
Journal of chromatographic science, 49(9), 657-664 (2012-05-16)
The determination of uranium at different stages of the recovery process as well as in seawater is important in its recovery study. A previous study developed a high-performance liquid chromatography (HPLC) method for uranium determination in seawater using α-hydroxy isobutyric
Jiao-Jiao Shangguan et al.
Applied biochemistry and biotechnology, 168(7), 1820-1833 (2012-10-12)
A 1,080-bp cDNA (CGMCC 2873) encoding of a cold-active lipase of Aspergillus fumigatus (AFL67) was cloned and expressed in Escherichia coli for the first time. The new lipase, AFL67, was one-step purified by 8.30 folds through Ni-NTA affinity chromatography with
Quan He et al.
Chirality, 24(2), 119-128 (2011-12-17)
The chiral discrimination mechanism in the resolution of sertraline with mandelic acid was investigated by examining the weak intermolecular interactions (such as hydrogen bond, CH/π, and van der Waals interactions) and molecular packing difference in crystal structures of the resulting
Yingzi Fu et al.
Analytical biochemistry, 421(1), 103-107 (2011-11-08)
A new chiral biosensor has been fabricated by immobilizing γ-globulin on gold nanoparticles modified glassy carbon electrodes, which could recognize and detect mandelic acid (MA) enantiomers. Differential pulse voltammetry, quartz crystal microbalance, ultraviolet-visible spectroscopy, and atomic force microscopy were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service