Skip to Content
Merck
All Photos(4)

Key Documents

151807

Sigma-Aldrich

Acetonitrile-d3

≥99.8 atom % D

Synonym(s):

Methyl-d3 cyanide, Trideuteroacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CN
CAS Number:
Molecular Weight:
44.07
Beilstein:
1740230
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99.8 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

3.0-16 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0200% water
water

refractive index

n20/D 1.341 (lit.)

bp

80.7 °C (lit.)

mp

-46 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1D3

InChI key

WEVYAHXRMPXWCK-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile-d3 (Trideuteroacetonitrile, CD3CN) is a deuterated NMR (Nuclear Magnetic Resonance) solvent. Molecular rotational friction coefficient and magnetic relaxation times (T1) for 14N and 2D of CD3CN have been determined. It participates as solvent during the synthesis of 2-phenylindole. It serves as solvent for 1H NMR (Proton Nuclear Magnetic Resonance) and 13C NMR (Carbon-13 Nuclear Magnetic Resonance) spectral studies.

Application

Acetonitrile-d3 has been used as a deuterated solvent for the dissolution of internal standard and sample in qNMR (Quantitative Nuclear Magnetic Resonance) quantification experiments. It has also been used as the deuterated solvent in the synthesis of N-methyl indolylfulgimide, which is studied using UV-pump-IR-probe spectroscopy.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Method development in quantitative NMR towards metrologically traceable organic certified reference materials used as 31P qNMR standards.
Weber M, et al.
Analytical and Bioanalytical Chemistry, 407(11), 91-98 (2015)
Synthesis and Structure of Homoleptic Iron Complex Bearing Pincer-Type 1, 2, 3-Triazol-5-ylidene.
Iwasaki H, et al.
Organic Chemistry Current Research, 6, 2161-0401 (2016)
Ultrafast Ring-Closure Reaction of Photochromic Indolylfulgimides Studied with UV-Pump- IR-Probe Spectroscopy
Koller.OF, et al.
The Journal of Physical Chemistry A, 112(2), 210-214 (2008)
Magnetic Resonance Studies of Anisotropic Molecular Rotation in Liquid Acetonitrile-d3.
Bopp TT.
J. Chem. Phys. , 47(9), 3621-3626 (1967)
[Cp*IrCl2]2-catalysed cyclization of 2-alkynylanilines into indoles.
Kumaran E and Leong WK.
Tetrahedron Letters, 55(40), 5495-5498 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service