Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetonitrile-d
3
Synonym(s):
Methyl-d
3
cyanide, Trideuteroacetonitrile
Linear Formula:
CD
3
CN
CAS Number:
2206-26-0
Molecular Weight:
44.07
EC Number:
218-616-5
Products
Product Category
solvents (5)
stable isotopes (3)
Brand
Sigma-Aldrich (7)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
151807
Acetonitrile-d
3
,
≥99.8 atom % D
Expand
View Pricing
366544
Acetonitrile-d
3
,
99.8 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
233323
Acetonitrile-d
3
,
"100%", 99.96 atom % D
Expand
View Pricing
1.02904
Acetonitrile-d
3
,
deuteration degree min. 99% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
569550
Acetonitrile-d
3
,
≥99.8 atom % D, anhydrous
Expand
View Pricing
233331
Acetonitrile-d
3
,
99.8 atom % D, contains 1 % (v/v) TMS
Expand
View Pricing
563102
Acetonitrile-d
3
,
96-97 atom % D, D
2
O 15-20 %
Expand
View Pricing