444324
Dichloromethane-d2
99.9 atom % D
Synonym(s):
Dideuteromethylenechloride, Methylene chloride-d2
About This Item
Recommended Products
isotopic purity
99.9 atom % D
Quality Level
Assay
99% (CP)
form
liquid
packaging
ampule of 0.6mL, 0.75mL, 1mL, 5g
glass bottle of 10g, 25g
technique(s)
NMR: suitable
impurities
≤0.0100% water
water
refractive index
n20/D 1.422 (lit.)
bp
40 °C (lit.)
mp
-97 °C (lit.)
density
1.362 g/mL at 25 °C (lit.)
mass shift
M+2
SMILES string
[2H]C([2H])(Cl)Cl
InChI
1S/CH2Cl2/c2-1-3/h1H2/i1D2
InChI key
YMWUJEATGCHHMB-DICFDUPASA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recommended products
accessory
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service