151599
Trichloroacetyl chloride
99%
Synonym(s):
2,2,2-Trichloroacetyl chloride, Trichloroacetic acid chloride, Trichloroacetochloride
About This Item
Recommended Products
vapor pressure
16 mmHg ( 20 °C)
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.470 (lit.)
bp
114-116 °C (lit.)
mp
−57 °C (lit.)
density
1.629 g/mL at 25 °C (lit.)
functional group
acyl chloride
chloro
SMILES string
ClC(=O)C(Cl)(Cl)Cl
InChI
1S/C2Cl4O/c3-1(7)2(4,5)6
InChI key
PVFOMCVHYWHZJE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
213.8 °F - open cup
Flash Point(C)
101 °C - open cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service