Skip to Content
Merck
All Photos(1)

Key Documents

367710

Sigma-Aldrich

Trichloroacetic anhydride

technical grade, 95%

Synonym(s):

(2,2,2-Trichloroacetyl) 2,2,2-trichloroacetate, Trichloroacetic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Cl3CCO)2O
CAS Number:
Molecular Weight:
308.76
Beilstein:
980355
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

95%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

139-141 °C/60 mmHg (lit.)

density

1.69 g/mL at 25 °C (lit.)

functional group

anhydride
chloro
ester

SMILES string

ClC(Cl)(Cl)C(=O)OC(=O)C(Cl)(Cl)Cl

InChI

1S/C4Cl6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10

InChI key

MEFKFJOEVLUFAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroacetic anhydride is reported to be an efficient derivatization reagent, since it produces stable derivatives having both a higher mass fragmentation pattern and lesser volatility than derivatives of trifluoroacetic anhydride (TFAA) or heptafluorobutyric anhydride (HFBA). The interaction of thymidine 5′-phosphate with trichloroacetic anhydride in dimethylformamide and acetonitrile in the presence of tertiary amines hs been studied.

Application

Trichloroacetic anhydride may be used as derivatization reagent for the following studies:
  • Simultaneous analysis of amphetamine, methamphetamine and 3,4-methylenedioxymethamphetamine (MDMA or Ecstasy) in urine samples by solid-phase extraction, derivatization and gas chromatography/mass spectrometry.
  • Analysis of halogenated amphetamines in brain tissue from rats by gas chromatography with electron-capture detection (GC-ECD).
  • Quantitative determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Czarny et al.
Journal of analytical toxicology, 13(5), 257-262 (1989-09-01)
An analytical procedure for methamphetamine and amphetamine has been developed using 4-carbethoxyhexafluorobutyryl chloride. This derivatizing reagent has all the previously reported advantages of trichloroacetic anhydride (TCAA) in that it produces stable derivatives having both a higher mass fragmentation pattern and
V S Bogachev et al.
Bioorganicheskaia khimiia, 29(1), 64-74 (2003-03-28)
The interaction of thymidine 5'-phosphate with trichloroacetic anhydride, trichloroacetyl chloride, and tribromoacetyl bromide was studied in dimethylformamide and acetonitrile in the presence of tertiary amines. The first two reactions gave the mixed anhydride of trichloroacetic and thymidylic acids (acyl phosphate)
D H Lau et al.
Journal of chromatography, 129, 329-338 (1976-12-22)
Six reagents-trichloroacetyl chloride, trichloroacetic anhydride, pentafluorobenzoyl chloride, heptafluorobutyryl chloride, heptafluorobutyric anhydride, and trifluoroacetic anhydride- were evaluated as potential derivatizing reagents for quantitating the metabolites of l-alpha-acetylmethadol (LAAM) -noracetylmethadol, dinoracetylmethadol, methadol, and normethadol-by electron capture gas-liquid chromatography. All of the reagents
B K Gan et al.
Journal of forensic sciences, 36(5), 1331-1341 (1991-09-01)
A rapid and effective solid-phase extraction procedure using Bond Elute Certify bonded silica sorbent cartridges was adopted to extract amphetamine, methamphetamine, and 3,4-methylenedioxymethamphetamine (MDMA or Ecstasy) from urine samples. The extract was derivatized with trichloroacetic anhydride prior to gas chromatography/mass
M L Owen et al.
Journal of pharmacological methods, 25(2), 147-155 (1991-04-01)
p-Chloro-alpha-fluoromethylphenylethylamine (fluoro-p-chloroamphetamine) (FpCA) has been shown in acute studies to be a less potent depletor of the neurotransmitter amine 5-hydroxytryptamine (5-HT) in brain than is p-chloroamphetamine (pCA). Gas chromatographic assay procedures for FpCA and PCA have been developed in our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service