Skip to Content
Merck
All Photos(1)

Key Documents

217328

Sigma-Aldrich

Trichloroacetyl isocyanate

96%

Synonym(s):

2,2,2-Trichloroacetyl isocyanate, alpha,alpha,alpha-Trichloroacetyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCONCO
CAS Number:
Molecular Weight:
188.40
Beilstein:
971201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.480 (lit.)

bp

80-85 °C/20 mmHg (lit.)

density

1.581 g/mL at 25 °C (lit.)

functional group

amine
chloro
isocyanate

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)C(=O)N=C=O

InChI

1S/C3Cl3NO2/c4-3(5,6)2(9)7-1-8

InChI key

GRNOZCCBOFGDCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Conformational stability and vibrational IR and Raman spectra of trichloroacetyl isocyanate has been investigated. Trichloroacetyl isocyanate is commonly employed as in situ derivatizing reagent for the 13C NMR studies of alcohols, phenols and amines. It undergoes 1,5-cycloaddition reaction with anhydro-2-deoxy-D-erythro- and -L-threo-pent-1-enitols and 1,5-anhydro-2-deoxy-D- and -L-arabino-hex-1-enitols to yield [2+2] and [4+2] cycloadducts.

Application

Trichloroacetyl isocyanate was used in catalytic one-pot dehydrative glycosylation of 1-hydroxy carbohydrate. It was also used as a reagent for the conversion of alcohols to carbamates†.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[2+ 2] Cycloaddition of trichloroacetyl isocyanate to glycals.
Chmielewski M and Kaluza Z.
Carbohydrate Research, 167, 143-152 (1987)
Tetrahedron Letters, 48, 65-65 (2007)
Hassan M Badawi et al.
Journal of molecular modeling, 8(2), 44-49 (2002-05-29)
The conformational stability and vibrational infrared and Raman spectra of trichloroacetyl isocyanate (CCl3CONCO) were investigated by ab initio MP2 and density functional B3LYP calculations using the 6-311++G** basis set. From the potential energy scans of the internal rotations in both
NMR spectral studies-XII: Trichloroacetyl isocyanate as an in situ derivatizing reagent for 13C NMR spectroscopy of alcohols, phenols and amines.
Bose AK and Srinivasan PR.
Tetrahedron, 31(24), 3025-3029 (1975)
Tatsuya Shirahata et al.
Carbohydrate research, 345(6), 740-749 (2010-03-09)
Efficient catalytic and stereoselective glycosylation was achieved by activating a glycosyl N-trichloroacetylcarbamate with a catalytic amount of Lewis acid in the presence of a glycosyl acceptor and 5A molecular sieves. Catalytic one-pot dehydrative glycosylation of a 1-hydroxy carbohydrate was achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service