Skip to Content
Merck
All Photos(1)

Key Documents

149187

Sigma-Aldrich

Phenyl salicylate

ReagentPlus®, 99%

Synonym(s):

Salol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
Beilstein:
393969
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid crystal

bp

172-173 °C/12 mmHg (lit.)

mp

41-43 °C (lit.)

SMILES string

Oc1ccccc1C(=O)Oc2ccccc2

InChI

1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H

InChI key

ZQBAKBUEJOMQEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Short-time dynamics of glass-forming liquids: Phenyl salicylate (salol) in bulk liquid, dilute solution, and confining geometries
Kalampounias, A. G., et al.
J. Chem. Phys. , 118(18), 8340-8349 (2003)
Application of phenyl salicylate-sepiolite systems as ultraviolet radiation filters
Hoyo, C. D., Vicente, M. A., & Rives, V.
Clay Minerals, 33(3), 467-474 (1998)
Different Polymorphic Modifications of Phenyl Salicylate
Davydova, N. A., Melnik, V. I., Nesprava, V. V., & Reznichenko, V. Y.
Ukrainian Journal of Physics, 57(2), 140-144 (2012)
M Niyaz Khan
Advances in colloid and interface science, 159(2), 160-179 (2010-08-03)
A new method, based upon semi-empirical kinetic approach, for the determination of ion exchange constant for ion exchange processes occurring between counterions at the cationic micellar surface is described in this review article. Basically, the method involves a reaction kinetic
M Paulí et al.
Acta odontologica latinoamericana : AOL, 10(1), 1-9 (1997-01-01)
The mutans group of streptococci is considered to play a key role in the etiology of dental caries. We have evaluated the ability of different substances to prevent dental plaque formation without affecting Streptococcus sobrinus viability. Viable organisms were detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service